Skip to Content
Merck
All Photos(2)

Key Documents

151939

Sigma-Aldrich

Methanol-OD

99.5 atom % D

Synonym(s):

mono-Deuteromethanol, Methan(ol-d), Methyl alcohol-OD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OD
CAS Number:
Molecular Weight:
33.05
Beilstein:
1730948
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

Quality Level

Assay

99% (CP)

form

liquid

expl. lim.

5.5-36.5 % (lit.)

technique(s)

NMR: suitable

impurities

≤0.0250% water
water

refractive index

n20/D 1.327 (lit.)

bp

65.5 °C (lit.)

mp

-99 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC

InChI

1S/CH4O/c1-2/h2H,1H3/i2D

InChI key

OKKJLVBELUTLKV-VMNATFBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methan(ol-d) (Methanol-OD, CH3OD) is a deuterated form of methanol. Dissociation of CH3OD adsorbed on the clean W(100) surface resulted in the formation of hydrogen, carbon and oxygen (β-CO). Additionally, carbon dioxide, water and methyl formate were obtained when similar dissociation was conducted over the W(100)-(5×1)C surface. The dissociation process was monitored by temperature-programmed reaction spectroscopy. Self-diffusion coefficient, shear viscosities and densities of liquid CH3OD have been determined by NMR spin-echo technique.

Application

Methanol-OD may have been used as solvent during cis-to-trans thermal isomerization of 1,3-diphenyltriazene using laser-induced optoacoustic calorimetry technique.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactions of methanol on W (100) and W (100)-(5? 1) C surfaces.
Ko EI, et al.
J. Catal., 62(2), 264-274 (1980)
Transport processes in compressed liquid methanol.
Jonas J and Akai JA.
J. Chem. Phys. , 66(11), 4946-4950 (1977)
A flash photolysis and optoacoustic calorimetry study of the cis?trans isomerization of 1, 3-diphenyltriazene
Scaiano.C.J
Journal of Photochemistry and Photobiology A: Chemistry, 62(1), 75-81 (1991)

Related Content

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

NMR spectroscopy elucidates molecular structure and purity via nuclear spin states in a strong magnetic field.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service