Skip to Content
Merck
All Photos(3)

Key Documents

150339

Sigma-Aldrich

2-Chloropyridine-3-carboxylic acid

99%

Synonym(s):

2-Chloronicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO2
CAS Number:
Molecular Weight:
157.55
Beilstein:
119023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

176-178 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cccnc1Cl

InChI

1S/C6H4ClNO2/c7-5-4(6(9)10)2-1-3-8-5/h1-3H,(H,9,10)

InChI key

IBRSSZOHCGUTHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloropyridine-3-carboxylic acid was used in the preparation of 2-pyridyl 2-chloropyridine-3-carboxylate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Synthesis of Heterocyclic Analogues of Thioflavones from Haloheteroaromatic Acids.
Bull. Korean Chem. Soc., 33(4), 1375-1378 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service