Skip to Content
Merck
All Photos(1)

Key Documents

14616

Sigma-Aldrich

Zirconium(IV) carbonate basic

≥40% ZrO2 basis

Synonym(s):

Zirconium hydroxycarbonate, [μ-[Carbonato(2-)-κO:κO′]]dihydroxydioxodizirconium

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

composition

ZrO2, ≥40%

reaction suitability

reagent type: catalyst
core: zirconium

application(s)

battery manufacturing

SMILES string

O[Zr](=O)OC(=O)O[Zr](O)=O

InChI

1S/CH2O3.2H2O.2O.2Zr/c2-1(3)4;;;;;;/h(H2,2,3,4);2*1H2;;;;/q;;;;;2*+2/p-4

InChI key

ACWOHNQUPJAZIT-UHFFFAOYSA-J

Application

Zirconium(IV) carbonate basic can be used as a catalyst:
  • For Meerwein–Ponndorf–Verley reduction of HMF and furfural
  • In the one-pot conversion of dihydroxyacetone into ethyl lactate

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service