Skip to Content
Merck
All Photos(1)

Key Documents

146056

Sigma-Aldrich

3-Nitrobenzyl alcohol

98%

Synonym(s):

NOBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
2044769
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

175-180 °C/3 mmHg (lit.)

mp

26-32 °C (lit.)

density

1.29 g/mL at 20 °C (lit.)

functional group

hydroxyl
nitro

SMILES string

OCc1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7NO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-4,9H,5H2

InChI key

CWNPOQFCIIFQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

3-Nitrobenzyl alcohol is the best substrate for cytosolic alcohol dehydrogenase.

Application

3-Nitrobenzyl alcohol was employed as matrix during fast-atom bombardment mass spectrometry. It was used in isolation of palmitylated peptide fragment from bovine rhodopsin and its characterization by mass spectrometry.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte Mallet et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(21), 7944-7953 (2015-04-14)
A series of 2,5-distyrylfuran derivatives bearing pentafluorophenyl- and cyanovinyl units have been synthesized for aggregation-induced emission (AIE). The effect of the type and extent of the supramolecular connections on the AIE of the furan derivatives were examined and correlated with
Harry J Sterling et al.
Journal of the American Society for Mass Spectrometry, 23(2), 191-200 (2011-12-14)
The effects of aqueous solution supercharging on the solution- and gas-phase structures of two protein complexes were investigated using traveling-wave ion mobility-mass spectrometry (TWIMS-MS). Low initial concentrations of m-nitrobenzyl alcohol (m-NBA) in the electrospray ionization (ESI) solution can effectively increase
S Hartmann et al.
Biological mass spectrometry, 23(6), 362-368 (1994-06-01)
Fast atom bombardment mass spectra were successfully recorded for intact glycosylphenolphthiocerol dimycocerosates (phenolic glycolipids, PGLs) from Mycobacterium kansasii, M. leprae, M. tuberculosis, M. marinum, M. bovis and M. haemophilum. Characteristic fragment ions from the loss of the oligosaccharide moiety and
Daniel D MacDougall et al.
Journal of molecular biology, 427(9), 1801-1818 (2014-10-14)
Ribosomal subunit joining is a key checkpoint in the bacterial translation initiation pathway during which initiation factors (IFs) regulate association of the 30S initiation complex (IC) with the 50S subunit to control formation of a 70S IC that can enter
D I Papac et al.
The Journal of biological chemistry, 267(24), 16889-16894 (1992-08-25)
Bovine rhodopsin has been reported to be S-palmitylated at cysteines 322 and 323 (Ovchinnikov, Y. A., Abdulaev, N. G., and Bogachuk, A.S. (1988) FEBS Lett. 230, 1-5). Using a combination of enzymatic and chemical cleavage techniques in conjunction with tandem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service