Skip to Content
Merck
All Photos(3)

Key Documents

136905

Sigma-Aldrich

4-Methoxybenzyl alcohol

98%

Synonym(s):

p-Anisyl alcohol, Anis alcohol, Anise alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2OH
CAS Number:
Molecular Weight:
138.16
Beilstein:
636654
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.544 (lit.)

bp

259 °C (lit.)

mp

22-25 °C (lit.)

solubility

alcohol: freely soluble
diethyl ether: freely soluble
water: insoluble

density

1.113 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

COc1ccc(CO)cc1

InChI

1S/C8H10O2/c1-10-8-4-2-7(6-9)3-5-8/h2-5,9H,6H2,1H3

InChI key

MSHFRERJPWKJFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Methoxybenzyl alcohol is commonly used as a reagent to protect hydroxyl groups on alcohols, and phenols. 4-Methoxybenzyl alcohol on condensation with the silanol groups of fumed silica nanoparticles yields modified silica nanoparticles. It undergoes catalytic photooxidation by flavin-zinc(II)-cyclen complex to yield the corresponding benzaldehyde.

Application

4-Methoxybenzyl alcohol was used to study the photocatalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria B Arce et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 11(6), 1032-1040 (2012-03-17)
The knowledge of photochemical kinetics in colloidal systems is important in understanding environmental photochemistry on dispersed solid surfaces. As model materials for the chemically sorbed organic compounds present in natural environments, modified silica nanoparticles (NPs) were obtained here by condensation
Radek Cibulka et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(24), 6224-6231 (2004-10-16)
Flavin-zinc(II)-cyclen 10 contains a covalently linked substrate binding site (zinc(II)-cyclen) and a chromophore unit (flavin). Upon irradiation, compound 10 effectively oxidizes 4-methoxybenzyl alcohol (11-OCH3) to the corresponding benzaldehyde both in water and in acetonitrile. In the presence of air, the
Selective photocatalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde in organic-free water in a continuous annular fixed bed reactor.
Loddo V, et al.
International Journal of Chemical Reactor Engineering, 5(1) (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service