Skip to Content
Merck
All Photos(1)

Key Documents

188263

Sigma-Aldrich

3-Methyl-2-nitrobenzyl alcohol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
167.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

48-50 °C (lit.)

functional group

hydroxyl
nitro

SMILES string

Cc1cccc(CO)c1[N+]([O-])=O

InChI

1S/C8H9NO3/c1-6-3-2-4-7(5-10)8(6)9(11)12/h2-4,10H,5H2,1H3

InChI key

NELPTBUSFQMYOB-UHFFFAOYSA-N

General description

3-Methyl-2-nitrobenzyl alcohol undergoes condensation with N,N-dimethylformamide dimethyl acetal followed by catalytic hydrogenation to yield 7-hydroxymethyl-indole.

Application

3-Methyl-2-nitrobenzyl alcohol was used as starting reagent in the synthesis of 7-formyl-indole.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple Synthesis of 7-Formyl-indole.
Uchil VR, et al.
Synthetic Communications, 36(8), 1051-1056 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service