142026
Benzoyleneurea
97%
Synonym(s):
2,4(1H,3H)-Quinazolinedione
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder
mp
300 °C (lit.)
solubility
ammonium hydroxide: soluble 10 mg/mL
DMF: soluble
alcohols: slightly soluble
SMILES string
O=C1NC(=O)c2ccccc2N1
InChI
1S/C8H6N2O2/c11-7-5-3-1-2-4-6(5)9-8(12)10-7/h1-4H,(H2,9,10,11,12)
InChI key
SDQJTWBNWQABLE-UHFFFAOYSA-N
Application
Benzoyleneurea scaffold was used in the synthesis of novel protein geranylgeranyltransferase-I inhibitors. It was used to study the mechanism of inactivation of chymotrypsin and other serine proteases by benzoxazinones.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analytical biochemistry, 211(1), 23-27 (1993-05-15)
A fluorometric high-performance liquid-chromatographic method is described for the determination of cyanate in human plasma. The method is based on the derivatization of cyanate with 2-aminobenzoic acid (anthranilic acid), leading to a stable cyclic fluorescent product, 2,4(1H,3H)-quinazolinedione. The fluorescent product
Nucleic acids research, 24(6), 1127-1135 (1996-03-15)
A major limitation in triple-helix formation arises from the weak energy of interaction between the third strand and the double-stranded target. We tried to increase the stacking interaction contribution within the third strand by extending the aromatic domain of thymine.
Proceedings of the National Academy of Sciences of the United States of America, 113(7), 1706-1713 (2016-01-23)
Mycobacterium tuberculosis (Mtb) infects one-third of the world's population and in 2013 accounted for 1.5 million deaths. Fluoroquinolone antibacterials, which target DNA gyrase, are critical agents used to halt the progression from multidrug-resistant tuberculosis to extensively resistant disease; however, fluoroquinolone
Chemical & pharmaceutical bulletin, 51(11), 1273-1282 (2003-11-06)
Potent, specific, chemically stable and non-peptide/small-molecular inhibitors of puromycin-sensitive aminopeptidase, such as 3-(2,6-diethylphenyl)-2,4(1H,3H)-quinazolinedione (PAQ-22, 5), were prepared by the structural development of a potent PSA inhibitor, 2-(2,6-diethylphenyl)-1,2,3,4-tetrahydroisoquinoline-1,3-dione (PIQ-22, 4). The design was carried out partly by applying electrostatic potential field
Journal of molecular evolution, 32, 282-288 (1991-01-01)
Previous attempts to produce nonenzymatic template-directed oligomerizations of activated pyrimidines on polypurine templates have been unsuccessful. The only efficient reactions are those where the template is composed primarily of pyrimidines, especially cytosine. Because molecular evolution requires that a synthesized daughter
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service