Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phenyl isothiocyanate
Synonym(s):
PITC, Phenyl isothiocyanate
Linear Formula:
C
6
H
5
NCS
CAS Number:
103-72-0
Molecular Weight:
135.19
EC Number:
203-138-1
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Puriss (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
139742
Phenyl isothiocyanate,
reagent grade, 98%
Expand
View Pricing
317861
Phenyl isothiocyanate,
99%, for protein sequencing
Expand
View Pricing
8.07028
Phenyl isothiocyanate,
for synthesis
Expand
View Pricing
P1034
Phenyl isothiocyanate,
Sigma Grade, 8.36 M, suitable for solid phase protein sequencing analysis, ≥99% (GC), liquid
Expand
View Pricing
78780
Phenyl isothiocyanate,
for HPLC derivatization, the detection of alcohols and amines, ≥99.0%
Expand
View Pricing