Skip to Content
Merck
All Photos(3)

Key Documents

135070

Sigma-Aldrich

Benzenesulfonic acid

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3H
CAS Number:
Molecular Weight:
158.18
Beilstein:
742513
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

solid

functional group

sulfonic acid

SMILES string

OS(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H,7,8,9)

InChI key

SRSXLGNVWSONIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzenesulfonic acid, also known as besylic acid, is a strong organic acid commonly used as an acid catalyst in various organic reactions, such as esterification, alkylation, and condensation reactions. It is also used as a sulfonating agent to introduce sulfonic acid (SO3H) groups into organic compounds, which can be useful in the synthesis of several dye intermediates.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramasubbu Ramani et al.
Biomacromolecules, 9(5), 1390-1397 (2008-04-19)
We present lamellar self-assembly of cationic poly(L-histidine) (PLH) stoichiometrically complexed with an anionic surfactant, dodecyl benzenesulfonic acid (DBSA), which allows a stabilized conformation reminiscent of polyproline type II (PPII) left-handed helices. Such a conformation has no intrapeptide hydrogen bonds, and
Wang-Ping Ting et al.
Journal of hazardous materials, 156(1-3), 421-427 (2008-02-05)
A new approach for promoting ferric reduction efficiency using a different electrochemical cell and the photoelectro-Fenton process has been developed. The use of UVA light and electric current as electron donors can efficiently initiate the Fenton reaction. Benzene sulfonic acid
Garry D Sisk et al.
Electrophoresis, 30(19), 3366-3371 (2009-09-04)
In this research, ion transfer across the interface between two immiscible electrolyte solutions (ITIES) was used as a method of detection in a CE separation system. This method allows for the electrochemical detection of ionic analytes that cannot be easily
Wenyi Huang et al.
ACS nano, 6(11), 10178-10185 (2012-10-27)
High-performance graphene nanopapers are prepared from an aqueous solution of functional graphenes with benzenesulfonic acid groups via covalent bonds. The formed hydrophobic graphene nanopapers showed the highest tensile strength of 360 MPa and Young's modulus of 102 GPa for samples
Wen Huang et al.
The Journal of organic chemistry, 73(17), 6845-6848 (2008-08-07)
A novel, convenient, and efficient method has been developed for selective synthesis of spirocycle, polycycle, and di- and tetrahydronaphthalene systems from aryl-substituted propargylic alcohols by FeCl3- or TsOH-catalyzed multiple activations of unsaturated C-C bonds and C-H bonds.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service