Skip to Content
Merck
All Photos(3)

Key Documents

448737

Sigma-Aldrich

Benzenesulfonic acid monohydrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3H · H2O
CAS Number:
Molecular Weight:
176.19
Beilstein:
4552655
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

mp

42-49 °C (lit.)

functional group

sulfonic acid

SMILES string

O.OS(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S.H2O/c7-10(8,9)6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);1H2

InChI key

MVIOINXPSFUJEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Benzenesulfonic acid monohydrate (PhSO3H. H2O) can be used as:
  • A catalyst to synthesize α,β-unsaturated amides via Pd-catalyzed selective aminocarbonylation of styrenes with nitroarenes.[1]
  • A reactant to prepare amino acid benzyl ester salts for the study of gelation in nonpolar solvents.[2]
  • A catalyst for the conversion of oxydipropionitrile (nitrile) to di-n-propyl-oxydipropionate (ester).[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mike Barbeck et al.
International journal of molecular sciences, 21(9) (2020-05-02)
Bioresorbable collagenous barrier membranes are used to prevent premature soft tissue ingrowth and to allow bone regeneration. For volume stable indications, only non-absorbable synthetic materials are available. This study investigates a new bioresorbable hydrofluoric acid (HF)-treated magnesium (Mg) mesh in
Arenesulfonic Acids as Catalysts in the Alcholysis of Nitriles to Esters
James F and Bryan W
The Journal of Organic Chemistry, 23(8), 1225-1227 (1958)
Palladium-Catalyzed Carbonylative Synthesis of α, β -Unsaturated Amides from Styrenes and Nitroarenes
Peng, Jin-Bao and Geng, Hui-Qing and Li, Da and Qi, Xinxin and Ying, Jun and Wu, Xiao-Feng
Organic Letters, 20(16), 4988-4993 (2018)
Preparation of Benzyl and p-Nitrobenzyl Esters of Amino Acids1
SHIELDS JE, et al.
The Journal of Organic Chemistry, 26(5), 1491-1494 (1961)
Karl H Schneider et al.
Acta biomaterialia, 116, 246-258 (2020-09-02)
Vascular grafts with a diameter of less than 6 mm are made from a variety of materials and techniques to provide alternatives to autologous vascular grafts. Decellularized materials have been proposed as a possible approach to create extracellular matrix (ECM) vascular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service