Skip to Content
Merck
All Photos(1)

Key Documents

129526

Sigma-Aldrich

2-Isopropylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1363322
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.05 mmHg ( 25 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.526 (lit.)

bp

212-213 °C (lit.)

mp

12-16 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccccc1O

InChI

1S/C9H12O/c1-7(2)8-5-3-4-6-9(8)10/h3-7,10H,1-2H3

InChI key

CRBJBYGJVIBWIY-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

General description

2-Isopropylphenol is catabolized via a broad-spectrum meta cleavage pathway[1].

Application

2-Isopropylphenol was used to study the ability of 2-isopropylphenol to activate estrogen receptor (ER), androgen receptor (AR), progesterone receptor (PR) and estrogen-related receptor (ERR)[2].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Li et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(1), 201-207 (2009-09-22)
Some phenols have been suspected to modulate the endocrine systems of wildlife and humans, but less is known about their interactions with different types of nuclear receptors. In this study, the ability of 2-tert-butylphenol, 2-isopropylphenol, 4-tert-octylphenol (4-t-OP), 2,4-dichlorophenol (2,4-DCP), 3,4-dichlorophenol
F Reichlin et al.
Applied and environmental microbiology, 60(12), 4587-4591 (1994-12-01)
Pseudomonas sp. strain HBP1 Prp grew on 2-isopropylphenol as the sole carbon and energy source with a maximal specific growth rate of 0.14 h-1 and transient accumulation of isobutyric acid. Oxygen uptake experiments with resting cells and enzyme assays with
Martin Barann et al.
Anesthesia and analgesia, 106(3), 846-857 (2008-02-23)
5-Hydroxytryptamine type 3 (5-HT3) receptors are excitatory ligand-gated ion channels which are involved in postoperative nausea and vomiting. They are depressed by the anesthetic propofol, which, in contrast, enhances the activity of inhibitory ligand-gated ion channels such as gamma-aminobutyric acid
Yongnian Ni et al.
Talanta, 76(3), 513-521 (2008-07-01)
The binding interaction of the pesticide Isoprocarb and its degradation product, sodium 2-isopropylphenate, with bovine serum albumin (BSA) was studied by spectrofluorimetry under simulated physiological conditions. Both Isoprocarb and sodium 2-isopropylphenate quenched the intrinsic fluorescence of BSA. This quenching proceeded
Kevin A Harvey et al.
Bioorganic & medicinal chemistry, 18(5), 1866-1874 (2010-02-16)
The present study describes the characterization and evaluation of novel anticancer conjugates, 2,6-diisopropylphenol-docosahexaenoate (PP-DHA), and its analogues including 2,4-diisopropylphenol-docosahexaenoate (DIPP-DHA), 2-isopropylphenol-docosahexaenoate (IPP-DHA), 2-cyclohexanephenol-docosahexaenoate (CHP-DHA) and phenol-docosahexaenoate (P-DHA) on breast cancer cell lines. Representative breast cancer cell lines, based on estrogen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service