Skip to Content
Merck
All Photos(1)

Key Documents

B97208

Sigma-Aldrich

2-tert-Butyl-4-methylphenol

99%

Synonym(s):

2-tert-Butyl-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(CH3)OH
CAS Number:
Molecular Weight:
164.24
Beilstein:
1817645
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

crystals

bp

244 °C (lit.)

mp

50-52 °C (lit.)

SMILES string

Cc1ccc(O)c(c1)C(C)(C)C

InChI

1S/C11H16O/c1-8-5-6-10(12)9(7-8)11(2,3)4/h5-7,12H,1-4H3

InChI key

IKEHOXWJQXIQAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Hirose et al.
Cancer research, 51(3), 824-827 (1991-02-01)
The combined effects of low doses of promoters or carcinogens on two-stage forestomach carcinogenesis were examined in rats pretreated with N-methyl-N'-nitro-N-nitrosoguanidine. Groups of 15 rats were given a single 150 mg/kg body weight intragastric dose of N-methyl-N'-nitro-N-nitrosoguanidine. Starting 1 week
P Lambelet et al.
Skin pharmacology : the official journal of the Skin Pharmacology Society, 1(2), 115-121 (1988-01-01)
Radical reactions of anthralin and its metabolites with skin have been studied by ESR spectroscopy. The influence of compounds which are known to suppress inflammation are described. The ESR spectra recorded during the reaction of anthralin with skin were essentially
Y Kurata et al.
Japanese journal of cancer research : Gann, 81(8), 754-759 (1990-08-01)
The urinary bladder tumor-promoting potentials of the phenolic antioxidants, 2-tert-butyl-4-methylphenol (TBMP), propylparaben, catechol, resorcinol and hydroquinone, which are structurally related to butylated hydroxyanisole (BHA), were investigated in 170 male F344 rats. The animals were initially given 0.05% N-butyl-N-(4-hydroxybutyl)nitrosamine (BBN) as
Shudong Zhang et al.
AAPS PharmSciTech, 20(2), 75-75 (2019-01-12)
Drugs with pH-dependent solubility that have poor water solubility can be identified in the drug discovery pipeline. Some of them have poor oral absorption, which can result in insufficient efficacy. Micro-environmental pH-modifying solid dispersion (micro pHm SD) is a promising
Xiaoxuan Zhou et al.
Biomaterials science, 5(8), 1588-1595 (2017-04-20)
Metastasis is the main reason for cancer-associated mortality, and accurate diagnostic imaging of metastases is critical for the clinical administration and tailoring personalized treatments for metastatic tumors. However, magnetic resonance imaging of metastases in the liver is impeded by its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service