Skip to Content
Merck
All Photos(1)

Key Documents

10930

Sigma-Aldrich

Arachidic acid

synthetic, ≥99.0% (GC)

Synonym(s):

Arachic acid, Arachidinic acid, Eicosanoic acid, Icosanoic acid, NSC 93983

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18COOH
CAS Number:
Molecular Weight:
312.53
Beilstein:
1788211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (GC)

form

solid

quality

synthetic

mp

74-76 °C (lit.)

solubility

H2O: insoluble
benzene: freely soluble
chloroform: freely soluble
cold water: very slightly soluble
diethyl ether: freely soluble
petroleum ether: freely soluble

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-19H2,1H3,(H,21,22)

InChI key

VKOBVWXKNCXXDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Arachidic acid was used to study the growth of cadmium sulfide crystals under arachidic acid monolayers[1]. It may be used to compose Langmuir-Blodgett (LB) films[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epitaxial growth of size-quantized cadmium sulfide crystals under arachidic acid monolayers.
Yang J, et al.
The Journal of Physical Chemistry, 99(15), 5500-5504 (1995)
Morphology of microphase separation in arachidic acid/cadmium arachidate Langmuir-Blodgett multilayers.
Kurnaz ML and Schwartz DK.
The Journal of Physical Chemistry, 100(26), 11113-11119 (1996)
Gregor Thomas Dahl et al.
Materials (Basel, Switzerland), 12(18) (2019-09-08)
Zirconia nanoceramics are interesting materials for numerous high-temperature applications. Because their beneficial properties are mainly governed by the crystal and microstructure, it is essential to understand and control these features. The use of co-stabilizing agents in the sol-gel synthesis of
Caiyan Lu et al.
Analytical chemistry, 83(1), 351-358 (2010-12-03)
An organic delta layer system made of alternating Langmuir-Blodgett multilayers of barium arachidate (AA) and barium dimyristoyl phosphatidate (DMPA) was constructed to elucidate the factors that control depth resolution in molecular depth profile experiments. More specifically, one or several bilayers
Gillian Groeger et al.
Journal of neurochemistry, 109(5), 1544-1554 (2009-04-07)
Reactive oxygen species (ROS) have traditionally been viewed as a toxic group of molecules; however, recent publications have shown that these molecules, including H(2)O(2), can also strongly promote cell survival. Even though the retina has a large capacity to produce

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service