Merck
  • N-glycosylation and topology of the human SLC26 family of anion transport membrane proteins.
N-glycosylation and topology of the human SLC26 family of anion transport membrane proteins.
American journal of physiology. Cell physiology (2014-03-22)
Jing Li, Fan Xia, Reinhart A F Reithmeier
ABSTRACT
The human solute carrier (SLC26) family of anion transporters consists of 10 members (SLCA1-11, SLCA10 being a pseudogene) that encode membrane proteins containing ~12 transmembrane (TM) segments with putative N-glycosylation sites (-NXS/T-) in extracellular loops and a COOH-terminal cytosolic STAS domain. All 10 members of the human SLC26 family, FLAG-tagged at the NH2 terminus, were transiently expressed in HEK-293 cells. While most proteins were observed to contain both high-mannose and complex oligosaccharides, SLC26A2 was mainly in the complex form, SLC26A4 in the high-mannose form, and SLC26A8 was not N-glycosylated. Mutation of the putative N-glycosylation sites showed that most members contain multiple N-glycosylation sites in the second extracytosolic (EC) loop, except SLC26A11, which was N-glycosylated in EC loop 4. Immunofluorescence staining of permeabilized cells localized the proteins to the plasma membrane and the endoplasmic reticulum, with SLC26A2 highly localized to the plasma membrane. N-glycosylation was not a necessary requirement for cell surface expression as the localization of nonglycosylated proteins was similar to their wild-type counterparts, although a lower level of cell-surface biotinylation was observed. No immunostaining of intact cells was observed for any SLC26 members, demonstrating that the NH2-terminal FLAG tag was located in the cytosol. Topological models of the SLC26 proteins that contain an even number of transmembrane segments with both the NH2 and COOH termini located in the cytosol and utilized N-glycosylation sites defining the positions of two EC loops are presented.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Boric acid-11B, ≥99 atom % 11B
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atp11c
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Sigma-Aldrich
Boric acid, SAJ first grade, ≥99.5%
Supelco
Benalaxyl-M, PESTANAL®, analytical standard
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Octaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Octaethylene glycol monododecyl ether, ≥98% (GC)
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
Sigma-Aldrich
Boric acid, ReagentPlus®, ≥99.5%
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Boric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%