Skip to Content
Merck
All Photos(1)

Key Documents

L0900000

Lysine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Lysine monohydrochloride, (S)-2,6-Diaminohexanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)4CH(NH2)CO2H·HCl
CAS Number:
Molecular Weight:
182.65
Beilstein:
3563889
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lysine

manufacturer/tradename

EDQM

mp

263 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC([C@@H](N)CCCCN)=O.[H]Cl

InChI

1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1

InChI key

BVHLGVCQOALMSV-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lysine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guntur Fibriansah et al.
Nature communications, 6, 6341-6341 (2015-02-24)
Dengue virus (DENV) infects ~400 million people annually. There is no licensed vaccine or therapeutic drug. Only a small fraction of the total DENV-specific antibodies in a naturally occurring dengue infection consists of highly neutralizing antibodies. Here we show that
Nicholas I Proellocks et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3103-3113 (2014-04-08)
The genomes of malaria parasites (Plasmodium spp.) contain a family of genes encoding proteins with a Plasmodium helical interspersed subtelomeric (PHIST) domain, most of which are predicted to be exported into the parasite-infected human red blood cell (iRBC). Here, using
Lunzhi Dai et al.
Nature chemical biology, 10(5), 365-370 (2014-04-01)
We report the identification of a new type of histone mark, lysine 2-hydroxyisobutyrylation (Khib), and identify the mark at 63 human and mouse histone Khib sites, including 27 unique lysine sites that are not known to be modified by lysine
Audrey Letourneau et al.
Nature, 508(7496), 345-350 (2014-04-18)
Trisomy 21 is the most frequent genetic cause of cognitive impairment. To assess the perturbations of gene expression in trisomy 21, and to eliminate the noise of genomic variability, we studied the transcriptome of fetal fibroblasts from a pair of
Chunaram Choudhary et al.
Nature reviews. Molecular cell biology, 15(8), 536-550 (2014-07-24)
Lysine acetylation is a conserved protein post-translational modification that links acetyl-coenzyme A metabolism and cellular signalling. Recent advances in the identification and quantification of lysine acetylation by mass spectrometry have increased our understanding of lysine acetylation, implicating it in many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service