Skip to Content
Merck
All Photos(1)

Documents

426148

Sigma-Aldrich

Boric acid-11B

≥99 atom % 11B

Synonym(s):

11B Labeled boric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H311BO3
CAS Number:
Molecular Weight:
62.03
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 11B

density

1.435 g/mL at 25 °C (lit.)

mass shift

depleted

SMILES string

O[11B](O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H/i1+0

InChI key

KGBXLFKZBHKPEV-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Stassi et al.
Analytical and bioanalytical chemistry, 409(10), 2615-2625 (2017-02-01)
An efficient way to increase the binding capability of microcantilever biosensors is here demonstrated by growing zinc oxide nanowires (ZnO NWs) on their active surface. A comprehensive evaluation of the chemical compatibility of ZnO NWs brought to the definition of
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed
Jing Li et al.
American journal of physiology. Cell physiology, 306(10), C943-C960 (2014-03-22)
The human solute carrier (SLC26) family of anion transporters consists of 10 members (SLCA1-11, SLCA10 being a pseudogene) that encode membrane proteins containing ~12 transmembrane (TM) segments with putative N-glycosylation sites (-NXS/T-) in extracellular loops and a COOH-terminal cytosolic STAS
Sunita A Ramesh et al.
Nature communications, 6, 7879-7879 (2015-07-30)
The non-protein amino acid, gamma-aminobutyric acid (GABA) rapidly accumulates in plant tissues in response to biotic and abiotic stress, and regulates plant growth. Until now it was not known whether GABA exerts its effects in plants through the regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service