Merck
  • Improved arsenic(III) adsorption by Al2O3 nanoparticles and H2O2: evidence of oxidation to arsenic(V) from X-ray absorption spectroscopy.
Improved arsenic(III) adsorption by Al2O3 nanoparticles and H2O2: evidence of oxidation to arsenic(V) from X-ray absorption spectroscopy.
Chemosphere (2014-07-30)
Linda Önnby, Prashanth Suresh Kumar, Kajsa G V Sigfridsson, Ola F Wendt, Stefan Carlson, Harald Kirsebom
ABSTRACT
We have investigated the oxidation of inorganic As(III) with H2O2 catalysed by Al2O3, using X-ray absorption near-edge structure and extended X-ray absorption fine structure spectroscopy. The effects of different reaction conditions (pH, time and initial H2O2 concentration) were also studied as were the kinetics of the oxidation reaction. We demonstrated that As(III) was oxidized to As(V) in the presence of H2O2 and Al2O3. Furthermore, all arsenic species found on the Al2O3 surface were in the As(V) state. The presence of both Al2O3 and H2O2 was necessary for oxidation of As(III) to take place within the period of time studied. The oxidation kinetics indicate a mechanism where reversible As(III) binding to the alumina surface is followed by irreversible oxidation by H2O2 leading to strongly bound As(V). Results from this study indicate that there is a surface-catalysed oxidation of As(III) on Al2O3 by H2O2, a reaction that can take place in nature and can be of help in the development of novel treatment systems for As(III) removal.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, CP
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, activated, neutral, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, acidic, Brockmann I, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh