Skip to Content
Merck
All Photos(2)

Documents

202606

Sigma-Aldrich

Aluminum oxide

99.997% trace metals basis

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
NA.23

Assay

99.997% trace metals basis

form

powder

mp

2040 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Siddiqi et al.
Dalton transactions (Cambridge, England : 2003), 44(32), 14349-14353 (2015-07-23)
A simple one-step and gram-scale synthesis of [Au5Mes5] from AuCl3 was developed, and this molecular precursor was used to generate Au nanoparticles on SiO2 and Al2O3. While [Au5Mes5] does not react with surface silanols and is only physisorbed, its incipient
Chien-Chih Lin et al.
Nanoscale, 5(17), 8090-8097 (2013-07-25)
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with
Joohoon Kang et al.
ACS nano, 9(4), 3596-3604 (2015-03-19)
Solution dispersions of two-dimensional (2D) black phosphorus (BP)--often referred to as phosphorene--are achieved by solvent exfoliation. These pristine, electronic-grade BP dispersions are produced with anhydrous organic solvents in a sealed-tip ultrasonication system, which circumvents BP degradation that would otherwise occur
Linda Önnby et al.
Chemosphere, 113, 151-157 (2014-07-30)
We have investigated the oxidation of inorganic As(III) with H2O2 catalysed by Al2O3, using X-ray absorption near-edge structure and extended X-ray absorption fine structure spectroscopy. The effects of different reaction conditions (pH, time and initial H2O2 concentration) were also studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service