Merck
  • Effectiveness of gentian violet and similar products commonly used to treat pyodermas.
Effectiveness of gentian violet and similar products commonly used to treat pyodermas.
Dermatologic clinics (2010-11-26)
Ricardo L Berrios, Jack L Arbiser
ABSTRACT
The term pyoderma encompasses a variety of distinct entities including impetigo (bullous and nonbullous), erysipelas, cellulitis, folliculitis, and staphylococcal scalded skin syndrome. Treatment of pyodermas centers around wound care and appropriate antibiotic selection. Triphenylmethane dyes, such as gentian violet, represent a unique group of compounds that act as antiseptics and have shown clinical efficacy as antibiotics in a variety of pyodermas, including those secondary to methicillin-resistant Staphylococcus aureus. Given their low cost, ease of application, and favorable side effect profile, triphenylmethanes must be considered legitimate treatment options for pyodermas, particularly in the face of continued and emerging bacterial resistance.
Sigma-Aldrich
Crystal Violet, JIS special grade
Sigma-Aldrich
Crystal Violet, SAJ first grade
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, S. No.: 785
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Gentian Violet solution
Sigma-Aldrich
Gram-Hucker′s stain solution (I)
Sigma-Aldrich
Gentian Violet, meets USP testing specifications