Skip to Content
Merck
All Photos(1)

Key Documents

12-0330

Sigma-Aldrich

Gentian Violet solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

composition

Gentian Violet, 1 % (w/v)

availability

available only in Japan

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

bacterial staining

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gram stain for Vibrio species.
Chia-Chang Kuo et al.
The American journal of emergency medicine, 31(5), 877-878 (2013-03-19)
Jayant S Raut et al.
Biofouling, 29(1), 87-96 (2012-12-12)
Biofilm-related infections caused by Candida albicans and associated drug resistant micro-organisms are serious problems for immunocompromised populations. Molecules which can prevent or remove biofilms are needed. Twenty-eight terpenoids of plant origin were analysed for their activity against growth, virulence attributes
Lise Crémet et al.
Diagnostic microbiology and infectious disease, 75(3), 252-255 (2013-01-15)
Biofilm formation seems to be a key factor in many bacterial infections, particularly those involving prosthetic implants or urinary catheters, where Escherichia coli is frequently involved. We have determined the ability to form biofilm in vitro of 34 E. coli
J X Zhang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(4), 737-744 (2013-01-12)
The adsorption of crystal violet dye from aqueous solutions onto an activated carbon prepared from peanut shells was analyzed in this study. The effects of particle size, initial concentration, time and temperature on crystal violet removal were studied in batch
Ziru Lian et al.
Journal of separation science, 36(5), 980-985 (2013-02-08)
A highly selective sample cleanup procedure combined with molecularly imprinted SPE was developed for the isolation of crystal violet from seawater and seafood samples. The molecularly imprinted polymer was prepared using crystal violet as the template molecule, methacrylic acid as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service