Skip to Content
Merck
All Photos(1)

Key Documents

05-6710

Sigma-Aldrich

Crystal Violet

JIS special grade

Synonym(s):

Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
Colour Index Number:
42555
Beilstein:
3580948
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

JIS special grade

availability

available only in Japan

mp

205 °C (dec.) (lit.)

antibiotic activity spectrum

fungi

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hector Alvarez et al.
PLoS genetics, 7(3), e1001356-e1001356 (2011-04-13)
Although a combination of genomic and epigenetic alterations are implicated in the multistep transformation of normal squamous esophageal epithelium to Barrett esophagus, dysplasia, and adenocarcinoma, the combinatorial effect of these changes is unknown. By integrating genome-wide DNA methylation, copy number
Tomoyuki Fujita et al.
Journal of the National Cancer Institute, 100(13), 940-949 (2008-06-26)
Neuroblastomas are characterized by hemizygous 1p deletions, suggesting that a tumor suppressor gene resides in this region. We previously mapped the smallest region of consistent deletion to a 2-Mb region of 1p36.31 that encodes 23 genes. Based on mutation analysis
Daniela Trisciuoglio et al.
Journal of the National Cancer Institute, 100(17), 1233-1246 (2008-08-30)
Candidaspongiolide (CAN), a novel polyketide from a marine sponge, is the active component of a mixture that was found to be potently cytotoxic in the National Cancer Institute's 60-cell-line screen. Effects of CAN on U251 glioma and HCT116 colorectal cancer
Gian Carlo Alghisi et al.
PloS one, 4(2), e4449-e4449 (2009-02-13)
Cilengitide is a high-affinity cyclic pentapeptdic alphaV integrin antagonist previously reported to suppress angiogenesis by inducing anoikis of endothelial cells adhering through alphaVbeta3/alphaVbeta5 integrins. Angiogenic endothelial cells express multiple integrins, in particular those of the beta1 family, and little is
Lene K Vestby et al.
BMC veterinary research, 5, 43-43 (2009-11-26)
Biofilm has been shown to be one way for Salmonella to persist in the feed factory environment. Matrix components, such as fimbriae and cellulose, have been suggested to play an important role in the survival of Salmonella in the environment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service