Skip to Content
Merck
All Photos(1)

Key Documents

32675

Sigma-Aldrich

Crystal Violet

S. No.: 785

Synonym(s):

Basic Violet 3, Gentian Violet, Hexamethylpararosaniline chloride, Methyl Violet 10B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C25H30N3Cl
CAS Number:
Molecular Weight:
407.98
Colour Index Number:
42555
Beilstein:
3580948
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02
Pricing and availability is not currently available.

Assay

≥96% (HPLC)

Quality Level

form

powder

color

S. No.: 785

mp

205 °C (dec.) (lit.)

solubility

water: 0.1%

density

1.190 g/cm3

suitability

complies for sensitivity

antibiotic activity spectrum

fungi

application(s)

diagnostic assay manufacturing
hematology
histology

Mode of action

cell membrane | interferes
enzyme | inhibits

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc(cc1)\C(c2ccc(cc2)N(C)C)=C3/C=C\C(C=C3)=[N+](/C)C

InChI

1S/C25H30N3.ClH/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6;/h7-18H,1-6H3;1H/q+1;/p-1

InChI key

ZXJXZNDDNMQXFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Crystal violet (CV) is a cationic tri-arylmethane dye, commercially used in ink and dyes. Organoclays based on CV are widely used in analytical studies.[1]

Application

CV was used to sensitize the photooxidation of cysteine to cysteic acid, as an end product hydrogen peroxide is also formed.[2]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of fast green and erythrosin-B to montmorillonite modified with crystal violet.
Rytwo G
Journal of Thermal Analysis and Calorimetry, 84 (1), 225-231 (2006)
FURTHER STUDIES ON THE CRYSTAL-VIOLET-SENSITIZED PHOTOOXIDATION OF CYSTEINE TO CYSTEIC ACID.
Gennari G and Cauzzo G.
Photochemistry and Photobiology, 20 (6), 497-500 (1974)
Teresa M Bernaciak et al.
Breast cancer research : BCR, 11(4), R52-R52 (2009-07-28)
Signal transducer and activator of transcription (STAT) 5b is a transcription factor involved in pro-proliferative and pro-survival signaling in a number of solid tumors, including breast cancer. The contribution of STAT5b to breast cancer cell motility has not been explored.
Tomoyuki Fujita et al.
Journal of the National Cancer Institute, 100(13), 940-949 (2008-06-26)
Neuroblastomas are characterized by hemizygous 1p deletions, suggesting that a tumor suppressor gene resides in this region. We previously mapped the smallest region of consistent deletion to a 2-Mb region of 1p36.31 that encodes 23 genes. Based on mutation analysis
Dai Miyazaki et al.
Nippon Ganka Gakkai zasshi, 117(4), 351-356 (2013-06-19)
To evaluate the efficacy of Gram-Fungiflora Y double staining for corneal scraping samples collected by impression in diagnosing infectious keratitis. Two hundred and thirteen eyes of 192 patients suspected of having infectious keratitis were retrospectively studied for the sensitivity and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service