Merck
  • Comparison of three methods to study biofilm formation by clinical strains of Escherichia coli.
Comparison of three methods to study biofilm formation by clinical strains of Escherichia coli.
Diagnostic microbiology and infectious disease (2013-01-15)
Lise Crémet, Stéphane Corvec, Eric Batard, Myriam Auger, Irene Lopez, Fabrice Pagniez, Sandie Dauvergne, Nathalie Caroff
ABSTRACT
Biofilm formation seems to be a key factor in many bacterial infections, particularly those involving prosthetic implants or urinary catheters, where Escherichia coli is frequently involved. We have determined the ability to form biofilm in vitro of 34 E. coli isolates by 3 different methods (crystal violet staining, BioFilm Ring Test®, and resazurin assay) and tried to correlate biofilm production with phylogenetic background and with the presence of different genes involved in biofilm synthesis. Only 3 isolates (including positive control E. coli ATCC 25922) were classified as strong biofilm producers (1B1, 1D, and 1B2 = control) by the 3 methods, 2 isolates by 2 different methods, and 5 additional isolates by only 1 method. All isolates possessed the csgA gene belonging to the csgABC operon encoding curli, and its regulator csgD. By contrast, only 76% possessed pgaA gene which is part of the pgaABCD operon encoding a polysaccharide adhesin. Interestingly, one of the strong biofilm producers did not harbor pgaA. In the second part, we have selected 5 specific isolates to study the impact of various experimental conditions on biofilm formation. For all these isolates, biofilm production was decreased in anaerobiosis and increased in LB medium compared with brain heart infusion medium, but at various degrees for the different isolates. These results underline the problems encountered in comparing the different published studies using various methods to study biofilm formation in vitro and the great need of standardization.
Sigma-Aldrich
Gentian Violet, meets USP testing specifications
Sigma-Aldrich
Gentian Violet solution
Sigma-Aldrich
Gram-Hucker′s stain solution (I)
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, indicator for the determination of the redox potential, S. No.: 785
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, JIS special grade
Sigma-Aldrich
Crystal Violet, SAJ first grade
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution