Skip to Content
Merck
All Photos(2)

Key Documents

57299-U

Supelco

SPME Fiber Assembly

greener alternative

DVB/CAR/PDMS, for use with manual holder or autosampler, Nitinol-core (NIT), 23 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)

Synonym(s):

DVB/CAR/PDMS SPME Fiber Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

Product Name

SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub

material

StableFlex fiber
notched gray hub

needle size

23 ga

packaging

pk of 3

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

2 cm

matrix active group

50/30µm DVB/CAR/PDMS
DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

Autosampler
for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

SPME fiber assemblies can be reused for up to 100 analyzes, or more, depending on the application and the care they are given. For reuse, simply condition with heat before and after every analysis. Solvent can be used for HPLC applications or when heat does not sufficiently clean the fiber. Each assembly has a color-coded or notched hub indicating the type of coating on the fiber. Smaller molecules are retained in the pores of the fibers containing adsorbents in the coating; e.g. carboxen, divinylbenzene particles.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Features and Benefits

Designed for use with Merlin Microseal sealing system; can be used with other septumless systems.

Other Notes

Appearance may vary. Darker spots may be visible on the fiber but will not affect performance.

Footnote

Coating bonded to a flexible fused silica core, yielding a more suitable coating on a less breakable fiber. There may be a slight difference in extraction selectivity compared to the same coating on a standard fused silica core. 2cm fiber assembly contains no spring.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pawliszyn, Janusz
Handbook of Solid Phase Microextraction, 410-410 (2009)
Pawliszyn, Janusz
Solid Phase Microextraction: Theory and Practice, 264-264 (1997)
Edited by Sue Ann Scheppers Wercinski
Solid Phase Microextraction: A Practical Guide, 257-257 (1999)
Pawliszyn, Janusz
Applications of Solid Phase Microextraction, 674-674 (1999)
Talon, R., Montel, M.
Applied SPME, 364-371 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service