Skip to Content
Merck
All Photos(3)

Documents

57328-U

Supelco

SPME Fiber Assembly

greener alternative

DVB/CAR/PDMS, for use with manual holder, Nitinol-core (NIT), 24 ga, 50 μm (DVB layer), 30 μm (CAR/PDMS layer)

Synonym(s):

StableFlex 1CM SPME FIBER PK3,MANUAL,50/30UM DVB/CARBOXEN-PD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115724
NACRES:
NB.21

product name

SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder

material

StableFlex fiber
plain gray hub

needle size

24 ga

packaging

pk of 3

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

30 μm (CAR/PDMS layer)
50 μm (DVB layer)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

DVB/CAR/PDMS coating

application(s)

food and beverages

compatibility

for analyte group flavors (volatiles and semivolatiles, C3-20) (MW 40-275)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Flavor Compounds

Other Notes

Appearance may vary. Darker spots may be visible on the fiber but will not affect performance.

Footnote

Coating bonded to a flexible fused silica core, yielding a more suitable coating on a less breakable fiber. There may be a slight difference in extraction selectivity compared to the same coating on a standard fused silica core.

Legal Information

StableFlex is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edgar Amavizca et al.
Scientific reports, 7, 41310-41310 (2017-02-02)
Remote effects (occurring without physical contact) of two plant growth-promoting bacteria (PGPB) Azospirillum brasilense Cd and Bacilus pumilus ES4 on growth of the green microalga Chlorella sorokiniana UTEX 2714 were studied. The two PGPB remotely enhanced the growth of the
Helena M Leal et al.
Scientific reports, 7(1), 17965-17965 (2017-12-23)
Anecdotes related to preferential mosquito bites are very common, but to date there is no complete explanation as to why one out of two people systematically receives more mosquito bites than the other when both are equally accessible. Here we
Odimar Z Zanardi et al.
Scientific reports, 8(1), 455-455 (2018-01-13)
Under laboratory conditions, mating activity in Asian citrus psyllid (ACP) started 4 days after emergence, peaked at day 7, and showed a clear window of activity starting 8 h into the photophase and extending through the first hour of the scotophase.
Sílvia M Rocha et al.
Journal of chromatography. A, 1252, 155-163 (2012-07-11)
Metabolomics represents an emerging issue that can aid in the diagnosis and/or prognosis of different diseases. Metabolomic study of urine is particularly interesting as it can be on the base of the developing of new faster and non-invasive methodologies. In
Sanja Risticevic et al.
Journal of chromatography. A, 1251, 208-218 (2012-07-10)
Metabolomics currently represents one of the fastest growing high-throughput molecular analysis platforms that refer to the simultaneous and unbiased analysis of metabolite pools constituting a particular biological system under investigation. In response to the ever increasing interest in development of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service