57294-U
SPME Fiber Assembly
PA, for use with autosampler, Nitinol-core (NIT), 23 ga, 85 μm (PA)
Synonym(s):
Polyacrylate SPME Fiber Assembly
About This Item
Recommended Products
Product Name
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
material
fused silica fiber
plain white hub
Quality Level
needle size
23 ga
packaging
pack of 3
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
df
85 μm (PA)
technique(s)
solid phase microextraction (SPME): suitable
fiber L
1 cm
matrix active group
PA coating
application(s)
food and beverages
compatibility
for analyte group polar semivolatiles (MW 80-300)
for use with autosampler
greener alternative category
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
Other Notes
Application
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
suitable for GC, application for SPMEsuitable for GC, application for SPMEOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service