52779-U
Supelclean Ultra 2400 solid phase extraction (SPE) Tubes
volume 1 mL, pkg of 108
Synonym(s):
Supelclean™ Ultra
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
Supelclean Ultra 2400, volume 1 mL, pkg of 108
product line
Supelclean™
Quality Level
composition
bed A, 120 mg
bed B, 100 mg (Z-Sep)
packaging
pkg of 108
technique(s)
solid phase extraction (SPE): suitable
volume
1 mL
matrix active group
C18 coating
Carbon coating
PSA coating
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
Supelclean™ PSA, Discovery® C18, and Spherical Carbon
This multi-bed solid phase extraction (SPE) cartridge was designed for the cleanup of extracts of difficult matrices such as dry commodities (tea, spices, coffee, etc.) prior to pesticide residue analysis, typically performed by GC/MSMS and LC/MS/MS. These types of samples can contain pigments and oils, and may not be sufficiently cleaned using a standard QuEChERS cleanup.
The top bed consists of a mixture of PSA/C18 and a graphitized, spherical carbon. PSA removes acidic interferences, C18 retains some hydrophobic interferences, and carbon removes pigments. The variety of carbon used in this cartridge was engineered to remove sufficient pigmentation while allowing for better recoveries of compounds with planar structures. The bottom bed of the cartridge contains Z-Sep, which is a zirconia-coated silica. Z-Sep removes oily residues, and provides additional pigment removal.
The cartridge can be used for cleanup of acetonitrile extracts to be analyzed by either GC or HPLC.
The top bed consists of a mixture of PSA/C18 and a graphitized, spherical carbon. PSA removes acidic interferences, C18 retains some hydrophobic interferences, and carbon removes pigments. The variety of carbon used in this cartridge was engineered to remove sufficient pigmentation while allowing for better recoveries of compounds with planar structures. The bottom bed of the cartridge contains Z-Sep, which is a zirconia-coated silica. Z-Sep removes oily residues, and provides additional pigment removal.
The cartridge can be used for cleanup of acetonitrile extracts to be analyzed by either GC or HPLC.
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
Supelclean is a trademark of Sigma-Aldrich Co. LLC
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service