Skip to Content
Merck
All Photos(2)

Key Documents

50994-U

Supelco

BIOshell Glycan (2.7 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

Product Name

BIOshell Glycan HPLC Columns, L × I.D. 15 cm × 2.1 mm, 2.7 μm particle size

material

stainless steel hardware (column)

Quality Level

Agency

suitable for USP L86

description

shell thickness (0.5 μm)
solid core (1.7 μm)

product line

BIOshell

feature

endcapped: no

analyte chemical class(es)

glycans

manufacturer/tradename

BIOshell

packaging

1 ea of

parameter

1000 bar max. pressure (14500 psi)
1000 bar pressure (14500 psi)
60 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

15 cm × 2.1 mm

surface area

135 m2/g

matrix

spherical silica particle platform
superficially porous particle

matrix active group

hydroxy, penta- phase

particle size

2.7 μm

pore size

90 Å

operating pH range

2-9

separation technique

hydrophilic interaction (HILIC)

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell Glycan HPLC Columns are specifically engineered to deliver fast, high resolution, reproducible glycan identification using HILIC chromatography. The Fused-Core columns provide improved resolution when compared to conventional LC methods thus allowing more reliable quantitation and confident identification of glycans.

View the BIOshell Glycan HPLC Columns Flyer

Application

BIOshell Glycan HPLC Columns are high-performance liquid chromatography columns with Fused-Core particle design. Their column chemistry is suitable for hydrophilic interactive liquid chromatography (HILIC) analysis of biomolecules like oligosaccharides.

Features and Benefits

BIOshell Glycan HPLC Columns are compatible both with UHPLC and HPLC systems. They are specifically engineered for the fast and high-resolution seperation of biomolecules. These columns operate at high flow rates and have 40% higher efficiency than fully porous particles of the same size. They are stable at high temperatures, which increases the throughput and enhances the peak shape.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service