Skip to Content
Merck
All Photos(2)

Key Documents

1.03728

Supelco

Water

suitable for UHPLC, LiChrosolv®

Synonym(s):

DI water, Deionized water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
MDL number:
UNSPSC Code:
41104213
NACRES:
NA.03
technique(s):
UHPLC: suitable
application(s):
general analytical
bp:
100 °C (lit.)
vapor pressure:
23 hPa ( 20 °C)
Pricing and availability is not currently available.

Product Name

Water, for UHPLC-MS LiChrosolv®

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
suitable for SM 4500 - NH3

Quality Level

reg. compliance

suitable for FDA C-010.02

vapor pressure

23 hPa ( 20 °C)

product line

LiChrosolv®

form

liquid

technique(s)

UHPLC: suitable

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)
0.0 °C (32.0 °F)

solubility

water: miscible

density

1.000 g/mL at 3.98 °C (lit.)

cation traces

Al: ≤5 ppb
Ca: ≤5 ppb
Fe: ≤5 ppb
K: ≤5 ppb
Mg: ≤10 ppb
Na: ≤50 ppb

application(s)

general analytical

storage temp.

2-30°C

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv® solvents exhibit high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them ideal for reproducible separations.

Features and Benefits

  • suitability tested and specified for UHPLC-MS and UHPLC-UV: for analytical flexibility
  • specified quality in positive and negative in positive and negative electrospray ionization (ESI) and atmospheric-pressure chemical ionization-mass spectrometry (APCI-MS) for lowest detection limits and confidence in analyses for all important MS modes (test 1)
    —ESI/APCI (+) < 2 ppb
    —ESI/APCI (-) < 10 ppb
  • lowest impurity profile: for interference-free baselines (test 2)
  • microfiltered through 0.2 μm filter (test 3) to provide:
    —a long lifespan to filters and mechanical parts of HPLC systems
    —reduced risk of column clogging
  • packed in borosilicate glass bottles for minimized metal ion contaminations
  • lowest levels of trace metal impurities for a minimum metal ion adduct formation
    —<5 ppb
  • lowest level of polyethylene glycol (PEG) impurities in UHPLC-MS solvent lineup (PEG S/N signal-to-noise-ratio < 50)

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

  1. Are there any products tested to ensure less than or equal to 1 ppb TOC? If not, what are the available options for water tested at the ppb level?

    1 answer
    1. Products tested to ensure less than or equal to 1 ppb TOC are not available. However, there are options for water that is tested to the ppb level. The items of potential interest include the following:

      1. 15333: less than or equal to 30 ppb TOC
      2. 03728: less than or equal to 10 ppb TOC
      3. 02699: less than or equal to 30 ppb TOC
      4. 03702: less than or equal to 30 ppb TOC
      The item that may be of most interest is product 1037281002, as the specification is less than or equal to 10 ppb for TOC.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service