Skip to Content
Merck
All Photos(1)

Key Documents

1.03726

Supelco

Methanol

99.9% (GC), suitable for UHPLC, LiChrosolv®

Synonym(s):

MeOH, Hydroxymethane, Carbinol, Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein:
1098229
MDL number:
UNSPSC Code:
41116105
EC Index Number:
200-659-6
NACRES:
NA.03
technique(s):
HPLC: suitable
LC/MS: suitable
UHPLC: suitable
application(s):
PFAS testing
bp:
64.7 °C (lit.)
vapor pressure:
128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

Product Name

Methanol, for UHPLC-MS LiChrosolv®

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for GB 5009.253-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

Quality Level

reg. compliance

suitable for FDA C-010.02

vapor density

1.11 (vs air)

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

product line

LiChrosolv®

Assay

99.9% (GC)

form

liquid

autoignition temp.

725 °F

expl. lim.

36 %

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC: suitable

refractive index

n20/D 1.329 (lit.)

kinematic viscosity

0.54-0.59 cSt(20 °C)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

SMILES string

CO

application(s)

PFAS testing

format

neat

storage temp.

2-30°C

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

UHPLC-MS LiChrosolv® solvents are designed to meet the highest requirements of UHPLC-MS in research and quality control, including proteomics and metabolomics and environmental, clinical, food or industrial testing applications

Application

Methanol is a high-purity solvent used as a mobile phase component in ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS) analysis.
The product is used as a solvent in studies such as the following:
  • high resolution mass spectrometric (HRMS) analysis of per- and polyfluoroalkyl substances (PFAS) in environmental water samples
  • UHPLC-MS/MS study of PFAS and/or perfluorooctane sulfonate (PFOS) in sewage sludge and soil samples

Features and Benefits

  • suitability tested and specified for UHPLC-MS and UHPLC-UV: for analytical flexibility
  • specified quality in positive and negative in positive and negative electrospray ionization (ESI) and atmospheric-pressure chemical ionization-mass spectrometry (APCI-MS) for lowest detection limits and confidence in analyses for all important MS modes (test 1)
—ESI/APCI (+) < 2 ppb
—ESI/APCI (-) < 10 ppb
  • lowest impurity profile: for interference-free baselines (test 2)
  • microfiltered through 0.2 μm filter (test 3) to provide:
—a long lifespan to filters and mechanical parts of HPLC systems
—reduced risk of column clogging
  • packed in borosilicate glass bottles for minimized metal ion contaminations
  • lowest levels of trace metal impurities for a minimum metal ion adduct formation
— <5 ppb
  • lowest level of polyethylene glycol (PEG) impurities in UHPLC-MS solvent lineup (PEG S/N signal-to-noise-ratio < 50)

Analysis Note

Evaporation Number: 1.9

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per-and polyfluoroalkyl substances in selected sewage sludge in Nigeria
Sindiku O, et al.
Chemosphere, 92(3), 329-335 (2013)
Qun Peng et al.
Scientific reports, 10(1), 3378-3378 (2020-02-27)
Acanthopanax trifoliatus (L.) Merr. (A. trifoliatus) belongs to the family Araliaceae, which is called "Le Cai", and is an indigenous plant to Guangdong Province that has been prevalently planted for years. A. trifoliatus is used in folk medicine and has
Are (fluorinated) ionic liquids relevant environmental contaminants? High-resolution mass spectrometric screening for per-and polyfluoroalkyl substances in environmental water samples led to the detection of a fluorinated ionic liquid
Neuwald IJ, et al.
Analytical and Bioanalytical Chemistry, 412(20), 4881-4892 (2020)
Electrodialytic per-and polyfluoroalkyl substances (PFASs) removal mechanism for contaminated soil
S?reng?rd M, et al.
Chemosphere, 232, 224-231 (2019)
Stabilization and solidification remediation of soil contaminated with poly-and perfluoroalkyl substances (PFASs)
S?reng?rd M, et al.
Journal of Hazardous Materials, 367, 639-646 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service