Skip to Content
Merck
All Photos(2)

Key Documents

50631-U

Supelco

Ascentis® Express 90 Å F5 (5 μm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Synonym(s):

Core-shell (SPP) Fused Core PFP HPLC column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L43

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

60 °C temp. range
600 bar max. pressure (9000 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 4.6 mm

surface area

90 m2/g

impurities

<5 ppm metals

matrix

Fused-Core particle platform
superficially porous particle

matrix active group

PFP (pentafluorophenyl) phase

particle size

5 μm

pore size

90 Å pore size

operating pH

2-8

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Ascentis® Express 5 μm F5 is a high-speed, high-performance liquid chromatography column based on the highly efficient Fused-Core® particle design. The bonded, endcapped, dimethylpentafluorophenyl-propylsilane stationary phase of Ascentis Express 5 μm F5 provides a stable, reversed-phase packing with electron-deficient phenyl rings due to the presence of electronegative fluorines. In addition to forming π-π and mildly steric interactions, F5 phases also retain compounds by polar interactions. As a result of having both polar and non-polar character, F5 phases can show dual-mode retention behavior, sometimes producing a “U-shaped” retention as a function of acetonitrile content of the mobile phase, with retention increasing at both low and high concentrations of ACN (reversed-phase and HILIC retention modes). Ascentis Express 5 μm F5 can be used for basic, acidic, or neutral compounds with alternate selectivity from C18.

Application


  • Liquid chromatographic determination of lumacaftor in the presence of ivacaftor and identification of five novel degradation products using high-performance liquid chromatography ion trap time-of-flight mass spectrometry.: This study employed the Ascentis® Express F5, 5 µm HPLC column to determine lumacaftor and its degradation products in the presence of ivacaftor. The application of high-performance liquid chromatography ion trap time-of-flight mass spectrometry provided high sensitivity and resolution, showcasing the column′s efficacy in complex sample matrices (Özcan et al., 2023).

  • Stability-indicating LC-MS/MS and LC-DAD methods for robust determination of tasimelteon and high resolution mass spectrometric identification of a novel degradation product.: This research highlights the use of the Ascentis® Express F5, 5 µm HPLC column in developing stability-indicating methods for tasimelteon. The column′s performance in both LC-MS/MS and LC-DAD methods ensured accurate and reliable identification of degradation products (Özcan et al., 2020).

  • Recent trends in ultra-fast HPLC: new generation superficially porous silica columns.: This review discusses advancements in HPLC technology, including the Ascentis® Express F5, 5 µm HPLC column. The article emphasizes the column′s capability in ultra-fast separations, providing enhanced efficiency and resolution in various analytical applications (Ali et al., 2012).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iltaf Shah et al.
The Journal of steroid biochemistry and molecular biology, 180, 118-128 (2018-02-13)
Research shows that immunoassay techniques are not the best choice for the estimation of vitamin D in human blood samples. The main reasons are that some immunoassays are not able to distinguish between 25-OHD3 and 25-OHD2 vitamin D metabolites. Furthermore
Maria A Khalikova et al.
Talanta, 130, 433-441 (2014-08-28)
The presented work describes the development of a simple, fast and effective on-line SPE-UHPLC-UV/vis method using fused core particle columns for extraction, separation and quantitative analysis of the nine illegal dyes, most frequently found in chilli-containing spices. The red dyes
Alex D Batista et al.
Talanta, 133, 142-149 (2014-12-02)
On-line sample pretreatment (clean-up and analyte preconcentration) is for the first time coupled to sequential injection chromatography. The approach combines anion-exchange solid-phase extraction and the highly effective pentafluorophenylpropyl (F5) fused-core particle column for separation of eight sulfonamide antibiotics with similar
Jan Soukup et al.
Journal of chromatography. A, 1374, 102-111 (2014-12-30)
Excess adsorption of water from aqueous acetonitrile mobile phases was investigated on 16 stationary phases using the frontal analysis method and coulometric Karl-Fischer titration. The stationary phases include silica gel and silica-bonded phases with different polarities, octadecyl and cholesterol, phenyl
Imran Ali et al.
Journal of separation science, 35(23), 3235-3249 (2012-11-28)
New generation columns, i.e. packed with superficially porous silica particles are available as trade names with following manufacturers: Halo, Ascentis Express, Proshell 120, Kinetex, Accucore, Sunshell, and Nucleoshell. These provide ultra-fast HPLC separations for a variety of compounds with moderate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service