Skip to Content
Merck
All Photos(1)

Documents

F8775

Sigma-Aldrich

Formaldehyde solution

for molecular biology, 36.5-38% in H2O

Synonym(s):

Formalin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCHO
CAS Number:
Molecular Weight:
30.03
Beilstein:
1209228
EC Number:
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.31

grade

for molecular biology

vapor density

1.03 (vs air)

vapor pressure

52 mmHg ( 37 °C)

form

liquid

autoignition temp.

572 °F

contains

10-15% methanol

concentration

36.5-38% in H2O

technique(s)

ChIP: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

density

1.09 g/mL at 25 °C (lit.)

suitability

suitable for (use in Formaldehyde-Agarose Gel Electrophoresis of RNA)

application(s)

general analytical
genomic analysis

storage temp.

15-25°C

SMILES string

[H]C([H])=O

InChI

1S/CH2O/c1-2/h1H2

InChI key

WSFSSNUMVMOOMR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Formaldehyde, a colorless, pungent-smelling chemical, finds extensive application in various biological research and laboratory settings. Primarily supplied as a 37% solution (formalin), its diverse uses range from tissue preservation and sterilization to acting as a denaturant in RNA gel electrophoresis. However, it′s crucial to acknowledge its potential health risks and handle it with appropriate precautions.Multifaceted Functions:
  • Preservative: Formaldehyde′s ability to inhibit microbial growth makes it an effective preservative in biological samples, preventing decay and ensuring their integrity for further analysis. This property finds application in medical laboratories, museums, and even embalming fluids.
  • Fixative: In histology, formaldehyde acts as a fixative, cross-linking proteins and preserving tissue structures for microscopic examination. This allows researchers to visualize anatomical details and cellular components with remarkable clarity.
  • Denaturant: In molecular biology techniques like formaldehyde-agarose gel electrophoresis, formaldehyde denatures RNA molecules, preventing them from forming secondary structures and enabling size-based separation. This technique proves invaluable for studying gene expression and RNA integrity.

Application

Formaldehyde solution has been used for fixing of cells and tissue sections. It has been used in the formaldehyde agarose gel electrophoresis. Formaldehyde is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA.

Features and Benefits

  • Versatile and adaptable for a wide variety of laboratory and research applications
  • Suitable for Molecular Biology and Biochemical applications

Analysis Note

Tested for use in formaldehyde-agarose gel electrophoresis of RNA.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 1 - STOT SE 3

Target Organs

Eyes,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

133.0 °F - closed cup

Flash Point(C)

56.11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farrell RE
RNA Methodologies: A Laboratory Guide for Isolation and Characterization (2010)
Lun Zhao et al.
Nature communications, 10(1), 3640-3640 (2019-08-15)
Insight into high-resolution three-dimensional genome organization and its effect on transcription remains largely elusive in plants. Here, using a long-read ChIA-PET approach, we map H3K4me3- and RNA polymerase II (RNAPII)-associated promoter-promoter interactions and H3K9me2-marked heterochromatin interactions at nucleotide/gene resolution in rice.
RNA-dependent RNA Polymerase Assay for Hepatitis E Virus.
Nair VP et al.
Bio-protocol null
Burry RW
Immunocytochemistry: A Practical Guide for Biomedical Research (2009)
Yanwei Sha et al.
Molecular genetics & genomic medicine, 7(8), e807-e807 (2019-07-04)
Asthenozoospermia (AZS), also known as asthenospermia, is characterized by reduced motility of ejaculated spermatozoa and is detected in more than 40% of infertile patients. Because the proportion of progressive spermatozoa in severe AZS is <1%, severe AZS is an urgent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service