Merck
  • On-line SPE-UHPLC method using fused core columns for extraction and separation of nine illegal dyes in chilli-containing spices.
On-line SPE-UHPLC method using fused core columns for extraction and separation of nine illegal dyes in chilli-containing spices.
Talanta (2014-08-28)
Maria A Khalikova, Dalibor Satínský, Tereza Smidrkalová, Petr Solich
ABSTRACT
The presented work describes the development of a simple, fast and effective on-line SPE-UHPLC-UV/vis method using fused core particle columns for extraction, separation and quantitative analysis of the nine illegal dyes, most frequently found in chilli-containing spices. The red dyes Sudan I-IV, Sudan Red 7B, Sudan Red G, Sudan Orange G, Para Red, and Methyl Red were separated and analyzed in less than 9 min without labor-consuming pretreatment procedure. The chromatographic separation was performed on Ascentis Express RP-Amide column with gradient elution using mixture of acetonitrile and water, as a mobile phase at a flow rate of 1.0 mL min(-1) and 55°C of temperature. As SPE sorbent for cleanup and pre-concentration of illegal dyes short guard fused core column Ascentis Express F5 was used. The applicability of proposed method was proven for three different chilli-containing commercial samples. Recoveries for all compounds were between 90% and 108% and relative standard deviation ranged from 1% to 4% for within- and from 2% to 6% for between-day. Limits of detection showed lower values than required by European Union regulations and were in the range of 3.3-10.3 µg L(-1) for standard solutions, 5.6-235.6 µg kg(-1) for chilli-containing spices.
Product Description
Supelco
Ascentis® Express F5, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 4.6 mm, pkg of 3 ea
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 4.6 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 3 mm, pkg of 3 ea
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 7.5 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Ascentis® Express F5, 5 μm Guard Cartridge, 5 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Ascentis® Express F5, 5 μm HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Ascentis® Express RP-Amide, 5 μm HPLC Column, 5 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 3 cm × 3 mm
Supelco
Ascentis® Express F5, 2 μm Guard Cartridge, 2 μm particle size, L × I.D. 5 mm × 2.1 mm, pkg of 3 ea
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 2.1 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 5 cm × 3 mm
Supelco
Ascentis® Express F5, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 7.5 cm × 3 mm