Skip to Content
Merck
All Photos(1)

Key Documents

Z724416

3D Biotek 3D Insert PCL scaffold

for 12 well plates, 6 inserts/ 12 well plate

Synonym(s):

3D, 3D Cell Culture, 3D scaffolds, Scaffolds, cell scaffolds

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22
Pricing and availability is not currently available.

material

(polycaprolactone (PCL))
clear polycaprolactone (PCL)
clear wells

description

polycaprolactone (PCL)

sterility

sterile

quality

6 inserts/ 12 well plate

feature

lid: no

packaging

pack of 6 ea (6 inserts supplied in 12 well plate)

manufacturer/tradename

3D Biotek PCL303012-6

well maximum volume

300 μm

working volume

300 μL

color

clear

pore size

569 μm pore size
65.6 % porosity (+/- 0.9)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

3D Biotek offers cutting edge 3D cell culture products and technology at affordable prices, that are easy and convenient to use our 3D 3D Insert cell culture scaffolds are:

  • Terminally sterilized by gamma irradiation and ready for use
  • Compatible with most of your current 2D assays
  • Easy for imaging. Cell growth can be monitored by light microscope, no need to purchase any additional sophisticated equipment.
Polycaprolactone (PCL) is a biodegradable polyester material that has been used in many FDA approved implants, drug delivery devices, suture, adhesion barrier and now is available in 3D Biotek′s revolutionary 3D Insert ! The PCL freeform fabrication has also been widely used in the fabrication of porous 3D scaffolds for tissue engineering research.

1.100% open porosity
The pores of the products are 100% open and interconnected, making it easy for cells to be seeded through out the porous scaffolds and the nutrient and cell metabolism waste to be exchanged.

2. Well defined pore size and porous structure

3D Biotek′s precision micro-fabrication technology produces well-defined porous structure and ensures the reproducibility of the porous structure from batch to batch.

3. Organic solvent free

Cytotoxic organic solvents, such as chloroform and methylene chloride, are often used in fabricating PCL scaffold. 3D Biotek′s precision micro-fabrication technology is a solvent free manufacturing process. Therefore, the PCL 3D Insert is free of organic solvent.

  • 12 well compatible
  • Fiber Diameter: 300u
  • Pore Size: 300u
  • 6 Inserts/Pack

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos E Caicedo-Carvajal et al.
Journal of tissue engineering, 2011, 362326-362326 (2011-11-11)
Isolation and amplification of primary lymphoma cells in vitro setting is technically and biologically challenging task. To optimize culture environment and mimic in vivo conditions, lymphoma cell lines were used as a test case and were grown in 3-dimension (3D)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service