Z724300
3D Biotek 3D Insert™ PS scaffold
for 96 well plates, 24 inserts/96 well plate
Synonym(s):
3D, 3D Cell Culture, 3D scaffolds, Scaffolds, cell scaffolds
About This Item
Recommended Products
material
clear polystyrene
clear wells
polystyrene
description
polystyrene (PS)
sterility
sterile
quality
24 inserts/96 well plate
packaging
pack of 24 ea (24 inserts supplied in 96 well plate)
manufacturer/tradename
3D Biotek PS152096-24
size
96 wells
color
clear
pore size
~200 μm (Fiber spacing)
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
- Terminally sterilized by gamma irradiation and ready for use
- Compatible with most of your current 2D assays
- Easy for imaging. Cell growth can be monitored by light microscope, no need to purchase any additional sophisticated equipment.
- 96 well compatible
- Fiber Diameter: 150u
- Pore Size: 200u
- 24 Inserts/Pack
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service