Skip to Content
Merck
All Photos(2)

Key Documents

Z687553

3D Biotek 3D Insert PS scaffold

for 24 well plates, 12 inserts/24 well plate

Synonym(s):

3D, 3D Cell Culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

polystyrene

description

polystyrene (PS)

sterility

sterile

quality

12 inserts/24 well plate

packaging

pack of 12 ea (12 inserts supplied in 24 well plate)

manufacturer/tradename

3D Biotek PS152024-12

fiber diam.

150 μm

size

24 well

color

clear

pore size

~200 μm (Fiber spacing)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

3D Biotek offers cutting edge 3D cell culture products and technology at affordable prices. Now it is the right time to take advantage of our promotional offer and try our products. You will be amazed by how easy and convenient to use our 3D Insert cell culture scaffolds. Our 3D Insert cell culture scaffolds are:

  • Terminally sterilized by gamma irradiation and ready for use
  • Compatible with most of your current 2D assays
  • Easy for imaging. Cell growth can be monitored by light microscope, no need to purchase any additional sophisticated equipment.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mayra Paolillo et al.
Cells, 8(11) (2019-11-27)
Metastatic spread is mainly sustained by cancer stem cells (CSC), a subpopulation of cancer cells that displays stemness features. CSC are thought to be derived from cancer cells that undergo epithelial to mesenchymal transition (EMT), thus acquiring resistance to anoikis
Carlos E Caicedo-Carvajal et al.
Journal of tissue engineering, 2011, 362326-362326 (2011-11-11)
Isolation and amplification of primary lymphoma cells in vitro setting is technically and biologically challenging task. To optimize culture environment and mimic in vivo conditions, lymphoma cell lines were used as a test case and were grown in 3-dimension (3D)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service