Skip to Content
Merck
All Photos(1)

Key Documents

V3765

Sigma-Aldrich

VU0255035 hydrate

≥98% (HPLC)

Synonym(s):

CID24768606, ML012, N-(3-oxo-3-(4-(pyridine-4-yl)piperazin-1-yl)propyl)benzo[c][1,2,5]thiadiazole-4-sulfonamide hydrate, N-[3-oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N6O3S2 · xH2O
CAS Number:
Molecular Weight:
432.52 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

O.O=C(CCNS(=O)(=O)c1cccc2nsnc12)N3CCN(CC3)c4ccncc4

InChI

1S/C18H20N6O3S2.H2O/c25-17(24-12-10-23(11-13-24)14-4-7-19-8-5-14)6-9-20-29(26,27)16-3-1-2-15-18(16)22-28-21-15;/h1-5,7-8,20H,6,9-13H2;1H2

InChI key

NURHGMDBQMBSQI-UHFFFAOYSA-N

Biochem/physiol Actions

VU0255035 is the first highly selective antagonist at the orthosteric site of the M1 receptor (75-fold selective for M1 relative to other muscarininc subtypes and devoid of activity at other GPCRs, ion channels, transporters and kinases).
VU0255035 is the first highly selective antagonist at the orthosteric site of the M1 receptor (75-fold selective for M1 relative to other muscarininc subtypes and devoid of activity at other GPCRs, ion channels, transporters and kinases). There are no highly selective M1 muscarinic receptor antagonists. The existing non-selective drugs do not permit direct evaluation of the role of M1 receptors in CNS fucntions and do not premit therapeutic targeting of M1 receptors in various disease states in which M1 receptors are implicated (epilepsy, Parkinson′s disease, attention and cognitive disorders, dystonia, etc).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service