Skip to Content
Merck
All Photos(1)

Key Documents

V4640

Sigma-Aldrich

VU0357017 monohydrochloride

≥98% (HPLC)

Synonym(s):

4-[[2-[(2-Methylbenzoyl)amino]ethyl]amino]-1-piperidinecarboxylic acid ethyl ester monohydrochloride, CID 25010775, ML071 hydrochloride, SID 56353039

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27N3O3 · HCl
CAS Number:
Molecular Weight:
369.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CCOC(=O)N1CCC(CC1)NCCNC(=O)c2ccccc2C

InChI

1S/C18H27N3O3.ClH/c1-3-24-18(23)21-12-8-15(9-13-21)19-10-11-20-17(22)16-7-5-4-6-14(16)2;/h4-7,15,19H,3,8-13H2,1-2H3,(H,20,22);1H

InChI key

XKJQVUIXSBOCPP-UHFFFAOYSA-N

Biochem/physiol Actions

VU0357017 is a subtype-selective M1 muscarinic acetylcholine allosteric agonist. VU0357017 has a potency of 200 nM and Achmax of 81% and had no activity at M2-M5 up to the highest concentrations tested and also had little or no detectable antagonist activity at any other mAChR subtype at concentrations over 2 orders of magnitude higher than those required to activate M1 or activity at a large panel of GPCRs, ion channels, and transporters. In contrast, TBPB inhibited responses to ACh at each of the other mAChR subtypes. VU0357017 was active in reversing cognitive deficits induced by scopolamine in a preclinical rodent model.

Features and Benefits

This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X H Chen et al.
Genetics and molecular research : GMR, 12(3), 2876-2885 (2013-09-26)
The purpose of this study was to determine whether interleukin-10 (IL-10) promoter polymorphisms are associated with leprosy or their subtypes in ethnic groups from southwest China. Genotyping using TaqMan® SNP Genotyping Master Mix and ABI 7500 real-time PCR system was
Shahnaz Sali et al.
Hepatitis monthly, 13(7), e8743-e8743 (2013-09-26)
Hepatitis B virus (HBV) is one of the most common chronic viral infections in the world. Iran has a low to intermediate HBV prevalence and approximately 1.5 million people are living with HBV infection. The impact of HBV in Iran
Masayuki Ikeno et al.
The International journal of oral & maxillofacial implants, 28(5), e283-e289 (2013-09-26)
The objective of this pilot study was to histologically evaluate bone regeneration using a self-assembling peptide hydrogel scaffold with recombinant human bone morphogenetic protein-2 (rhBMP-2) in a rabbit calvaria model. Five adult New Zealand White rabbits were used for the
Andreia N Horácio et al.
PloS one, 8(9), e73704-e73704 (2013-09-26)
In Portugal, pneumococcal conjugate vaccines have been administered to children outside of the national immunization plan since 2001. We determined the serotype and antimicrobial susceptibility of 1265 isolates responsible for adult invasive pneumococcal infections (IPD) between 2009 and 2011 and
Carolin Gall et al.
Frontiers in human neuroscience, 7, 584-584 (2013-09-26)
Patients with visual field defects after visual pathway lesion may experience reduced vision-related quality of life (vrQoL). It has not been clarified how vrQoL impairments contribute to vision-related mental distress. One hundred and eight subjects with visual field defects caused

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service