Skip to Content
Merck
All Photos(1)

Key Documents

SML0255

Sigma-Aldrich

4-DAMP

≥98% (HPLC)

Synonym(s):

1,1-Dimethyl-4-diphenylacetoxypiperidinium iodide, 4-DAMP methiodide, 4-Diphenylacetoxy-N-methylpiperidine methiodide, 4-[(Diphenylacetyl)oxy]-1,1-dimethyl-piperidinium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26INO2
CAS Number:
Molecular Weight:
451.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥15 mg/mL

storage temp.

room temp

SMILES string

[I-].C[N+]1(C)CCC(CC1)OC(=O)C(c2ccccc2)c3ccccc3

InChI

1S/C21H26NO2.HI/c1-22(2)15-13-19(14-16-22)24-21(23)20(17-9-5-3-6-10-17)18-11-7-4-8-12-18;/h3-12,19-20H,13-16H2,1-2H3;1H/q+1;/p-1

InChI key

WWJHRSCUAQPFQO-UHFFFAOYSA-M

Biochem/physiol Actions

4-DAMP is a M3 preferring cholinergic receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shohei Yamaguchi et al.
Food chemistry, 276, 376-382 (2018-11-10)
Our previous results (Nakamura et al., 2013, 2016) indicated that acetylcholine (ACh) in orally administered foods exerts antihypertensive effects. Eggplants (Solanum melongena) contain abundant ACh (Horiuchi et al., 2003), and their food functionality was discovered, using spontaneously hypertensive rats, by
Qiang Liu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(3), 907-919 (2017-11-28)
Overactive bladder associated with bladder outlet obstruction (BOO) is a highly prevalent condition, which is usually treated with antimuscarinics. However, the potential effects of antimuscarinics on the structure and function of bladder have not been investigated thus far. Sprague-Dawley(R) rats
Hyun Jung Kim et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 43(5), 1790-1802 (2017-10-20)
Magnolia officinalis Rehder and EH Wilson (M. officinalis) are traditional Chinese medicines widely used for gastrointestinal (GI) tract motility disorder in Asian countries. We investigated the effects of an ethanol extract of M. officinalis (MOE) on the pacemaker potentials of
Run-Qing Xue et al.
Journal of cellular and molecular medicine, 21(1), 58-71 (2016-08-06)
Mitochondrial dynamics-fission and fusion-are associated with ischaemic heart disease (IHD). This study explored the protective effect of vagal nerve stimulation (VNS) against isoproterenol (ISO)-induced myocardial ischaemia in a rat model and tested whether VNS plays a role in preventing disorders
Mojtaba Mohseni et al.
Veterinary research forum : an international quarterly journal, 8(2), 115-120 (2017-08-09)
Displacement of the abomasum is a common disease of the gastrointestinal tract in dairy cattle. Abomasal displacement has been associated with abomasal hypomotility. Therefore, it is necessary to identify effective therapeutic agents that stimulate abomasal motility in cattle. Yarrow (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service