Skip to Content
Merck
All Photos(1)

Key Documents

T4393

Sigma-Aldrich

Thrombin from human plasma

lyophilized powder, 1500-3500 NIH units/mg protein (E1%/280, 18.3), suitable for cell culture

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human plasma

Quality Level

sterility

sterile

form

lyophilized powder

specific activity

1500-3500 NIH units/mg protein (E1%/280, 18.3)

technique(s)

cell culture | mammalian: suitable

impurities

HIV, hepatitis B and hepatitis C, tested negative

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... F2(2147)

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is produced from the proteolytic cleavage of inactive prothrombin in the liver. The prothrombin gene is mapped to human chromosome 11p11.2. It comprises of A and B catalytic domain, recognition domain and insertion loops. The active site residues comprise the catalytic tetrad, (histidine 57, aspartate 102, serine 195 and serine 214).

Application

Thrombin from human plasma has been used:
  • as a medium supplement for the pre-treatment of endothelial cell culture prior to confocal microscopy and enzyme linked immunosorbent assay (ELISA)
  • in the gelatinization of mesenchymal stem cells (MSCs) for preparing fibrin–MSC construct
  • for screening serine protease inhibitor, OGTI from frog skin secretion

Biochem/physiol Actions

The main function of thrombin is the cleavage of fibrinogen to fibrin, to assist stable clot formation. High levels of thrombin elicit neurotoxicity in dopaminergic neurons and contributes to the progression of Parkinson′s disease. A wide range of mutations in the prothrombin gene contributes to its deficiency resulting in coagulation disorders like dysprothrombinemia and hypoprothrombinemia. Altered thrombin levels modulates the coagulation pathway in multiple sclerosis. Patients with coronary artery disease (CAD) show elevated levels of thrombin. Thrombin accumulation in neurofibrillary tangles in the brain may contribute to the aggregation of τ protein and pathophysiology of Alzheimer disease.
Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard

Reconstitution

When reconstituted with 1 mL water, vial contains stated activity in 0.15 M sodium chloride and 0.05 M sodium citrate, pH 6.5.

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A small trypsin inhibitor from the frog of Odorrana grahami
Li J, et al.
Biochimie, 90(9), 1356-1361 (2008)
Richard C Rogers et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 318(6), R1068-R1077 (2020-04-23)
Severe trauma can produce a postinjury "metabolic self-destruction" characterized by catabolic metabolism and hyperglycemia. The severity of the hyperglycemia is highly correlated with posttrauma morbidity and mortality. Although no mechanism has been posited to connect severe trauma with a loss
Endothelial cell processing and alternatively spliced transcripts of factor VIII: potential implications for coagulation cascades and pulmonary hypertension
Shovlin CL, et al.
PLoS ONE, 5(2), e9154-e9154 (2010)
Thrombin generation correlates with disease duration in multiple sclerosis (MS): Novel insights into the MS-associated prothrombotic state
Parsons MEM, et al.
Multiple Sclerosis Journal, 3(4) (2017)
Thrombin and prothrombin are expressed by neurons and glial cells and accumulate in neurofibrillary tangles in Alzheimer disease brain
Arai T, et al.
Journal of Neuropathology and Experimental Neurology, 65(1), 19-25 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service