Skip to Content
Merck
All Photos(1)

Key Documents

T1063

Sigma-Aldrich

Thrombin from human plasma

lyophilized powder, ≥2800 NIH units/mg protein (E1%/280, 18.3)

Synonym(s):

Factor IIa

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥2800 NIH units/mg protein (E1%/280, 18.3)

mol wt

37.4 kDa

impurities

HIV, hepatitis B and hepatitis C, tested negative

UniProt accession no.

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Gene Information

human ... F2(2147)

Looking for similar products? Visit Product Comparison Guide

General description

Thrombin is the final coagulation protease in regard to hemostasis, promoting both procoagulant and anticoagulant effects.

Application

Thrombin is used for site specific cleavage of recombinant fusion proteins containing an accessible thrombin recognition site for removal of affinity tags. Thrombin has been used in a study to evaluate the in vitro effect of low molecular weight heparin on thrombin generation in cirrhotic patients at different stages of liver disease.

Biochem/physiol Actions

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.

Unit Definition

Activity is expressed in NIH units obtained by direct comparison to a NIH Thrombin Reference Standard

Physical form

Lyophilized from 0.02 M Bis/Tris buffer, pH 6.5, 0.15 M NaCl and 0.1% PEG-8000

Analysis Note

The NIH assay procedure uses 0.2 ml diluted plasma (1:1 with saline) as a substrate and 0.1ml of thrombin sample (stabilized in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15-25 seconds are used for determining thrombin concentrations.

Other Notes

View more information on thrombin at www.sigma-aldrich.com/enzymeexplorer.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong Feng Gao et al.
Nature protocols, 15(2), 316-337 (2020-01-10)
Current visual biosensing methods, including colorimetric-based, fluorescence-based and chemiluminescence-based methods, are inappropriate for the hundreds of millions of people affected by color blindness and color weakness. Compared with these available methods, a droplet motion-based strategy might be a promising protocol
M Senzolo et al.
Journal of thrombosis and haemostasis : JTH, 10(9), 1823-1829 (2012-06-21)
Cirrhotic patients may present thrombotic complications that warrant anticoagulant therapy. However, the efficacy of low-molecular-weight heparin (LMWH) in this clinical setting is still unclear. To evaluate the in vitro effect of LMWH on thrombin generation (TG) in cirrhotic patients at
D David et al.
British journal of haematology, 113(3), 604-615 (2001-05-31)
In haemophilia A, the functional defect at the molecular level of most factor VIII (FVIII) missense mutations remains unknown. Site-directed mutagenesis of B domain-deleted FVIII cDNA (FVIIISQ) was used to introduce two mutations associated with severe cross-reacting material (CRM)-negative (FVIII-C329S)
Sooyeon Kim et al.
RSC advances, 8(37), 20822-20828 (2018-06-06)
We propose a new sensor design that maximizes fluorescence contrast, inspired by whispering-gallery mode lasing (WGM). Aptamer-modified glass microspheres (cf. 1-38 μm) and thrombin are used as a model sensory cavity and target protein, respectively. Two types of microsphere are
Otilia Obreja et al.
Experimental dermatology, 15(1), 58-65 (2005-12-21)
Activation of protease-activated receptors (PAR) can induce vasodilation (VD) and increase of vascular permeability either directly by stimulating endothelial cells or indirectly via activation of nociceptors and subsequent release of neuropeptides (neurogenic inflammation). We aimed to estimate the relative contribution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service