Skip to Content
Merck
All Photos(1)

Key Documents

L2023

Sigma-Aldrich

Leupeptin trifluoroacetate salt

powder, ≥90% (HPLC)

Synonym(s):

Acetyl-Leu-Leu-Arg-al trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38N6O4 · xC2HF3O2
CAS Number:
Molecular Weight:
426.55 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Leupeptin trifluoroacetate salt, ≥90% (HPLC), microbial

biological source

microbial

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

H2O: 10 mM (Solutions are stable only a few hours. Stock solutions are stable up to 6 months at −20 °C.)

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC(C)C[C@H](NC(C)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCNC(N)=N)C=O

InChI

1S/C20H38N6O4.C2HF3O2/c1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;3-2(4,5)1(6)7/h11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);(H,6,7)/t15-,16-,17-;/m0./s1

InChI key

WUHGBZVQELGOMH-FRKSIBALSA-N

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM.
Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Analysis Note

Leupeptin gives multiple peaks on HPLC because of the formation of tautomeric isomers in solution. Majority of contaminating peptide is racemized leupeptin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Manzano-Lopez et al.
PloS one, 17(2), e0263617-e0263617 (2022-02-11)
Intracellular trafficking through the secretory organelles depends on transient interactions between cargo proteins and transport machinery. Cytosolic coat protein complexes capture specific luminal cargo proteins for incorporation into transport vesicles by interacting with them indirectly through a transmembrane adaptor or
Maryam Amirazodi et al.
Iranian journal of basic medical sciences, 25(2), 254-262 (2022-06-04)
High-intensity interval training (HIIT) is a shape of interval training that provides ameliorated athletic capacity and has a good effect on health. Resveratrol is a natural polyphenol abundant in grapes and red wine and has been demonstrated to apply various
John Cherrington Carmen et al.
Frontiers in microbiology, 2, 1-1 (2011-07-01)
The regulation of mitochondrial permeability, a key event in the initiation of apoptosis is governed by the opposing actions of the pro- and anti-apoptotic members of the BCL2-family of proteins. The BCL2-family can be classified further based on the number
N Korn et al.
Poultry science, 81(7), 1077-1085 (2002-08-07)
A monoclonal antibody (4C7/2E4) that binds to an antigen located on the midpiece of turkey spermatozoa has been produced and characterized. The antibody was also reactive to antigens on the midpiece of chicken, Japanese quail, and bobwhite quail spermatozoa but
Alexandra Weyrich et al.
Biochemistry and cell biology = Biochimie et biologie cellulaire, 86(5), 380-385 (2008-10-17)
It is controversial whether DNA methylation plays a functional role in Drosophila. We have studied testis DNA of Drosophila melanogaster Meigen, 1830 with antisera against 5-methylcytosine (5mC) and found no evidence for the presence of significant amounts of 5mC. Reactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service