Skip to Content
Merck
All Photos(3)

Key Documents

T2573

Sigma-Aldrich

Troglitazone

≥98% (HPLC), powder, PPARγ agonist

Synonym(s):

(±)-5-[4-[(6-Hydroxy-2,5,7,8-tetramethylchroman-2-yl)methoxy]benzyl]-2,4-thiazolidinedione, CS-045

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO5S
CAS Number:
Molecular Weight:
441.54
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Troglitazone, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: 20 mg/mL

SMILES string

Cc1c(C)c2OC(C)(CCc2c(C)c1O)COc3ccc(CC4SC(=O)NC4=O)cc3

InChI

1S/C24H27NO5S/c1-13-14(2)21-18(15(3)20(13)26)9-10-24(4,30-21)12-29-17-7-5-16(6-8-17)11-19-22(27)25-23(28)31-19/h5-8,19,26H,9-12H2,1-4H3,(H,25,27,28)

InChI key

GXPHKUHSUJUWKP-UHFFFAOYSA-N

Gene Information

General description

Troglitazone is used to treat type 2 diabetes. It is associated with liver injury. Troglitazone functions as an inducer of cytochrome3A.

Application

Troglitazone has been used as a reagent in adipocyte differentiation medium.

Biochem/physiol Actions

PPARγ agonist; anti-diabetic thiazolidinedione (TZD) with anti-inflammatory and anti-tumor activity; induces apoptosis via a p53 pathway.

Features and Benefits

This compound is featured on the Nuclear Receptors (PPARs) and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Studies using troglitazone have reported that it can increase AMP-activated protein kinase (pAMPK) and phosphorylated acetyl-CoA carboxylase in isolated rat extensor digitorum longus muscles1. Furthermore, analysis of troglitazone-mediated autophagy is reported to correlate with the stimulation of AMPK α2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reprogramming of Human Cord Blood CD34+ Cells into Induced MSCs
Hematopoietic Differentiation of Human Pluripotent Stem Cells, 91-101 (2015)
Troglitazone hepatotoxicity: are we getting closer to understanding idiosyncratic liver injury?
Jaeschke H
Toxicological Sciences, 97(1), 1-3 (2007)
Registered report: IDH mutation impairs histone demethylation and results in a block to cell differentiation
Richarson AD, et al.
eLife, 5, e10860-e10860 (2016)
The Practice of Medicinal Chemistry (2015)
Phil June Park et al.
International journal of molecular sciences, 20(8) (2019-04-18)
Adiponectin (APN), released mainly from adipose tissue, is a well-known homeostatic factor for regulating glucose levels, lipid metabolism, and insulin sensitivity. A recent study showed that human hair follicles express APN receptors and the presence of APN-mediated hair growth signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service