Skip to Content
Merck
All Photos(1)

Key Documents

SML2701

Sigma-Aldrich

CBA

≥98% (HPLC)

Synonym(s):

4-Chloro-2-[[2-(2-chlorophenoxy)acetyl]amino]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11Cl2NO4
CAS Number:
Molecular Weight:
340.16
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)OCC(=O)Nc2c(ccc(c2)Cl)C(=O)O

InChI

1S/C15H11Cl2NO4/c16-9-5-6-10(15(20)21)12(7-9)18-14(19)8-22-13-4-2-1-3-11(13)17/h1-7H,8H2,(H,18,19)(H,20,21)

InChI key

CVQCJPCMPGKEDH-UHFFFAOYSA-N

Biochem/physiol Actions

CBA (Compound 5; 4-Chloro-2-[[2-(2-chlorophenoxy)acetyl]amino]benzoic acid) is a cell penetrant, potent and selective TRPM4 (Transient receptor potential melastatin member 4) inhibitor that shows not significant activity against TRPM5, TRPM7, TRPM8, TRPV1, TRPV3, TRPV6. It rescues functional expression of mutant A432T TRPM4 in cells. CBA (Compound 5) exhibits neuroprotective activity against glutamate-induced neurodegeneration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijo Cherian Ozhathil et al.
British journal of pharmacology, 175(12), 2504-2519 (2018-03-27)
TRPM4 is a calcium-activated non-selective cation channel expressed in many tissues and implicated in several diseases, and has not yet been validated as a therapeutic target due to the lack of potent and selective inhibitors. We sought to discover a
Beatrice Bianchi et al.
Molecular brain, 11(1), 41-41 (2018-07-13)
Transient receptor potential melastatin member 4 (TRPM4), a Ca2+-activated nonselective cation channel, has been found to mediate cell membrane depolarization in immune response, insulin secretion, cardiovascular diseases, and cancer. In murine experimental autoimmune encephalomyelitis (EAE), TRPM4 deletion and administration of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service