Skip to Content
Merck
All Photos(1)

Documents

78314

Sigma-Aldrich

Ammonium formate solution

BioUltra, 10 M in H2O

Synonym(s):

Ammonium salt of formic acid, ammonium formate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONH4
CAS Number:
Molecular Weight:
63.06
MDL number:
UNSPSC Code:
12352106
eCl@ss:
39021903
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

form

liquid

concentration

10 M in H2O

impurities

insoluble matter, passes filter test

pH

6.0-8.0 (25 °C, 10 M in H2O)

solubility

H2O: 10 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

10 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.04

SMILES string

[NH4+].[O-]C=O

InChI

1S/CH2O2.H3N/c2-1-3;/h1H,(H,2,3);1H3

InChI key

VZTDIZULWFCMLS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium formate is readily available, stable and nontoxic in nature. It can be used in conjugation with palladium on carbon (Pd-C) or Raney-nickel catalysts.

Application

Ammonium formate has been used as a constituent in pre-mixed eluents for forming gradient during quantification of diuron by liquid chromatography-mass spectrometry (LC-MS).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of phototrophic stream biofilms under stress: comparing traditional and novel ecotoxicological endpoints after exposure to diuron
Sgier L, et al.
Frontiers in Microbiology, 9, 2974-2974 (2018)
Nadja S Sieber-Ruckstuhl et al.
Scientific reports, 9(1), 6015-6015 (2019-04-14)
Glucocorticoids (GCs) are critical regulators of metabolic control in mammals and their aberrant function has been linked to several pathologies. GCs are widely used in human and veterinary clinical practice as potent anti-inflammatory and immune suppressive agents. Dyslipidaemia is a
A general procedure for mild and rapid reduction of aliphatic and aromatic nitro compounds using ammonium formate as a catalytic hydrogen transfer agent.
Ram S and Ehrenkaufer RE.
Tetrahedron Letters, 25(32), 3415-3418 (1984)
Anne M Filppula et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(7), 1202-1209 (2014-04-10)
Previous studies have shown that several protein kinase inhibitors are time-dependent inhibitors of cytochrome P450 (CYP) 3A. We screened 14 kinase inhibitors for time-dependent inhibition of CYP2C8 and CYP3A. Amodiaquine N-deethylation and midazolam 1'-hydroxylation were used as marker reactions for
Todd A Lydic et al.
Methods (San Diego, Calif.), 87, 83-95 (2015-04-25)
There is an increasing recognition of the role that cancer cell derived exosomes play in intercellular signaling upon fusion or uptake with a target cell, including immune system evasion, tumor growth and metastasis. To date, however, although exosomal membrane and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service