Skip to Content
Merck
All Photos(1)

Key Documents

SML0872

Sigma-Aldrich

MJN110

≥98% (HPLC)

Synonym(s):

2,5-dioxopyrrolidin-1-yl 4-(bis(4-chlorophenyl)methyl)piperazine-1-carboxylate, Cravatt Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21Cl2N3O4
CAS Number:
Molecular Weight:
462.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

O=C(ON1C(CCC1=O)=O)N2CCN(C(C3=CC=C(Cl)C=C3)C4=CC=C(Cl)C=C4)CC2

InChI

1S/C22H21Cl2N3O4/c23-17-5-1-15(2-6-17)21(16-3-7-18(24)8-4-16)25-11-13-26(14-12-25)22(30)31-27-19(28)9-10-20(27)29/h1-8,21H,9-14H2

InChI key

BEADRWVIFHOSGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

MJN110 has been used as a monoacylglycerol lipase (MAGL) inhibitor to study its effect on aggressive grooming in rats. It has also been used to inhibit lipid droplet formation.

Biochem/physiol Actions

MJN110 is a potent selective inhibitor of MAGL, the enzyme predominantly responsible for the degradation of the endocannabinoid 2-arachidonoylglycerol (2-AG) with >10,000 selectivity over FAAH, the hydrolase that degrades the endocannabinoid anandamide (AEA). MJN110 inhibits rat, mouse and human MAGL with IC50 values ranging from < 100 nM in rat to an IC50 of ~1 nM with 10- and 100-fold selectivity over closely related ABHD6, a serine hydrolase that acts as an alternative hydrolase of 2-AG, and LYPLA1/2 in human PC3 cells. MJN110 showed potent anti-hyperalgesic activity in a rat model of diabetic neuropathy, showing a therapeutic potential for treating diabetes chronic pain.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoacylglycerol lipase inhibition alters social behavior in male and female rats after post-weaning social isolation
Fontenot J, et al.
Behavioural Brain Research, 341, 146-153 (2018)
Micah J Niphakis et al.
ACS chemical neuroscience, 4(9), 1322-1332 (2013-06-05)
Monoacylglycerol lipase (MAGL) is a principal metabolic enzyme responsible for hydrolyzing the endogenous cannabinoid (endocannabinoid) 2-arachidonoylglycerol (2-AG). Selective inhibitors of MAGL offer valuable probes to further understand the enzyme's function in biological systems and may lead to drugs for treating
Jenny L Wilkerson et al.
The Journal of pharmacology and experimental therapeutics, 357(1), 145-156 (2016-01-23)
Serious clinical liabilities associated with the prescription of opiates for pain control include constipation, respiratory depression, pruritus, tolerance, abuse, and addiction. A recognized strategy to circumvent these side effects is to combine opioids with other antinociceptive agents. The combination of
Jae Won Chang et al.
ACS chemical biology, 8(7), 1590-1599 (2013-05-25)
Serine hydrolases are one of the largest and most diverse enzyme classes in Nature. Inhibitors of serine hydrolases are used to treat many diseases, including obesity, diabetes, cognitive dementia, and bacterial and viral infections. Nonetheless, the majority of the 200+
Elizabeth A Fucich et al.
Journal of neurophysiology, 123(2), 707-717 (2020-01-09)
Our previous work showed that lateral fluid percussion injury to the sensorimotor cortex (SMC) of anesthetized rats increased neuronal synaptic hyperexcitability in layer 5 (L5) neurons in ex vivo brain slices 10 days postinjury. Furthermore, endocannabinoid (EC) degradation inhibition via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service