Skip to Content
Merck
All Photos(1)

Key Documents

SML2732

Sigma-Aldrich

DO34

≥98% (HPLC)

Synonym(s):

(2-Benzyl-4-{[(2-methyl-2-propanyl)oxy]carbonyl}piperazinyl){4-[(4-trifluoromethoxy)phenyl]-1H-1,2,3-triazol-1-yl}methanone, 3-(Phenylmethyl)-4-[[4-[4-(trifluoromethoxy)phenyl]-1H-1,2,3-triazol-1-yl]carbonyl]-1-piperazinecarboxylic acid, 1,1-dimethylethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28F3N5O4
CAS Number:
Molecular Weight:
531.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N1CCN(CC1CC2=CC=CC=C2)C(OC(C)(C)C)=O)N3N=NC(C4=CC=C(C=C4)OC(F)(F)F)=C3

Biochem/physiol Actions

DO34 is a brain-penetrant, selective and highly potent diacylglycerol lipase inhibitor (recom human DAGLα/β pIC50 = 8.2/8.1 by SAG hydrolysis; mouse brain DAGLα/β pIC50 = 9.1-9.3/8.6 by ABPP ReDiMe) with detectable off-target activity toward only ABHD6 & PLA2G7 among all mouse brain serine hydrolases and little affinity toward cannabinoid receptors CB1/2. DO34 blocks depolarization-induced suppression of excitation (DSE IC50 = 0.18 μM) and inhibition (100% DSI blockage at 1 μM) in mouse cerebellar and hippocampal slices ex vivo, and attenuate LPS-induced neuroinflammatory responses by lowering brain 2-AG & PGE2 level in mice in vivo (50 mg/kg, i.p).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carol A Gianessi et al.
Addiction biology, 25(3), e12768-e12768 (2019-05-06)
Individuals with alcohol use disorder exhibit compulsive habitual behaviors that are thought to be, in part, a consequence of chronic and persistent use of alcohol. The endocannabinoid system plays a critical role in habit learning and in ethanol self-administration, but
Jayme R McReynolds et al.
Biological psychiatry, 84(2), 85-94 (2017-11-05)
Clinical reports suggest that rather than directly driving cocaine use, stress may create a biological context within which other triggers for drug use become more potent. We hypothesize that stress-induced increases in corticosterone "set the stage" for relapse by promoting
Jenny L Wilkerson et al.
The Journal of pharmacology and experimental therapeutics, 363(3), 394-401 (2017-10-04)
Diacylglycerol lipase (DAGL) α and β, the major biosynthetic enzymes of the endogenous cannabinoid (endocannabinoid) 2-arachidonylglycerol (2-AG), are highly expressed in the nervous system and immune system, respectively. Genetic deletion or pharmacological inhibition of DAGL-β protects against lipopolysaccharide (LPS)-induced inflammatory
Daisuke Ogasawara et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(1), 26-33 (2015-12-17)
Diacylglycerol lipases (DAGLα and DAGLβ) convert diacylglycerol to the endocannabinoid 2-arachidonoylglycerol. Our understanding of DAGL function has been hindered by a lack of chemical probes that can perturb these enzymes in vivo. Here, we report a set of centrally active
Hui Deng et al.
Journal of medicinal chemistry, 60(1), 428-440 (2016-12-20)
Triazole ureas constitute a versatile class of irreversible inhibitors that target serine hydrolases in both cells and animal models. We have previously reported that triazole ureas can act as selective and CNS-active inhibitors for diacylglycerol lipases (DAGLs), enzymes responsible for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service