SML3763
HSL-IN-1
≥97% (NMR)
Synonym(s):
B-[5-Chloro-2-[[[6-[4-(trifluoromethyl)phenoxy]-3-pyridinyl]carbonyl]amino]phenyl]boronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C19H13BClF3N2O4
CAS Number:
Molecular Weight:
436.58
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21
Recommended Products
Quality Level
Assay
≥97% (NMR)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
HSL-IN-1 is an orally available, potent, and selective inhibitor of Hormone-sensitive lipase (HSL) that exhibits an antilipolytic effect on rats.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The dysfunction of hormone-sensitive lipase induces lipid deposition and reprogramming of nutrient metabolism in fish
The British Journal of Nutrition, 130(4), 588-603 (2023)
Omega-6 highly unsaturated fatty acids in Leydig cells facilitate male sex hormone production
Communications biology, 5(1), 1001-1001 (2022)
Identification of a novel hormone sensitive lipase inhibitor with a reduced potential of reactive metabolites formation
Bioorganic & Medicinal Chemistry, 25(7), 2234-2243 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service