Skip to Content
Merck
All Photos(1)

Key Documents

SML0197

Sigma-Aldrich

4E1RCat

≥97% (HPLC)

Synonym(s):

4-[(3E)-3-[[5-(4-nitrophenyl)furan-2-yl]methylidene]-2-oxo-5-phenylpyrrol-1-yl]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H18N2O6
CAS Number:
Molecular Weight:
478.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

color

faint brown to dark brown-red

solubility

DMSO: ≥5 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(cc1)N2C(=O)\C(=C\c3ccc(o3)-c4ccc(cc4)[N+]([O-])=O)C=C2c5ccccc5

InChI

1S/C28H18N2O6/c31-27-21(16-24-14-15-26(36-24)19-6-12-23(13-7-19)30(34)35)17-25(18-4-2-1-3-5-18)29(27)22-10-8-20(9-11-22)28(32)33/h1-17H,(H,32,33)/b21-16+

InChI key

BBQRBOIMSKMFFO-LTGZKZEYSA-N

Application

4E1RCat was used to inhibit new protein synthesis in transformed primary chicken embryo fibroblasts.

Biochem/physiol Actions

4E1RCat ilnhibits the eIF4F translation initiation complex by binding to IF4E b and inhibiting the interaction between IF4E:eIF4G (IC50 = 3.2 μM). In in vitro translation experiments using MDA-MB-231 cells, 4E1RCat dose dependently inhibited cap-dependent translation, led to a decrease in polysome and 80S ribosomal subunits and reduced levels of the eIF4F-dependant proteins Mcl-1 and c-Myc.
4E1RCat is an inhibitor of translation initiation complex eIF4F

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the MAPKAPs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guoning Liao et al.
PloS one, 8(6), e68190-e68190 (2013-07-11)
DIAPH1 is a formin protein which promotes actin polymerization, stabilizes microtubules and consequently is involved in cytoskeleton dynamics, cell migration and differentiation. In contrast to the relatively well-understood signaling cascades that regulate DIAPH1 activity, its spatial regulation of biogenesis is
Shensi Shen et al.
Nature communications, 10(1), 5713-5713 (2019-12-18)
Cancer persister cells tolerate anticancer drugs and serve as the founders of acquired resistance and cancer relapse. Here we show that a subpopulation of BRAFV600 mutant melanoma cells that tolerates exposure to BRAF and MEK inhibitors undergoes a reversible remodelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service