Skip to Content
Merck
All Photos(1)

Key Documents

324517

Sigma-Aldrich

eIF4E/eIF4G Interaction Inhibitor, 4EGI-1

The eIF4E/eIF4G Interaction Inhibitor, 4EGI-1, also referenced under CAS 315706-13-9, controls the biological activity of eIF4E/eIF4G interaction. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

eIF4E/eIF4G Interaction Inhibitor, 4EGI-1, 2-((4-(3,4-Dichlorophenyl)-thiazol-2-ylhydrazono)-3-(2-nitrophenyl))propionic acid, eIF4F Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12Cl2N4O4S
CAS Number:
Molecular Weight:
451.28
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (sum of two isomers, HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C18H12Cl2N4O4S/c19-12-6-5-10(7-13(12)20)15-9-29-18(21-15)23-22-14(17(25)26)8-11-3-1-2-4-16(11)24(27)28/h1-7,9H,8H2,(H,21,23)(H,25,26)/b22-14+

InChI key

KFRKRECSIYXARE-HYARGMPZSA-N

General description

A cell-permeable hydrazone compound that reversibly binds eukaryotic translation initiation factor 4E (eIF4E; KD = 25 µM) and disrupts eIF4E/eIF4G, but not eIF4E/4E-BP1, complex formation. Reported to downregulate the protein, but not mRNA, levels of c-Myc and Bcl-xL in Jurkat cells (8 h), leading to apoptotic cell death after prolonged exposure (24 h, IC50 = ~ 25 µM). Also available as a 25 mM solution in DMSO (Cat. No. 324519).
A cell-permeable hydrazone compound that reversibly binds eukaryotic translation initiation factor 4E (eIF4E; KD = 25 µM) and disrupts eIF4E/eIF4G, but not eIF4E/4E-BP1, complex formation. Reported to downregulate the protein, but not mRNA, levels of c-Myc and Bcl-xL in Jurkat cells (8 hrs), leading to apoptotic cell death after prolonged exposure (24 hrs, IC50 ~25 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Moerke, N. J., et al. 2007. Cell128, 257.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

677.1 °F - (calculated)

Flash Point(C)

358.4 °C - (calculated)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xinna Li et al.
Aging cell, 21(9), e13685-e13685 (2022-08-06)
Glycosylphosphatidylinositol-specific phospholipase D1 (GPLD1) hydrolyzes inositol phosphate linkages in proteins anchored to the cell membrane. Mice overexpressing GPLD1 show enhanced neurogenesis and cognition. Snell dwarf (DW) and growth hormone receptor knockout (GKO) mice show delays in age-dependent cognitive decline. We
Gerald C Tiu et al.
Developmental cell, 56(14), 2089-2102 (2021-07-10)
In ribosomopathies, perturbed expression of ribosome components leads to tissue-specific phenotypes. What accounts for such tissue-selective manifestations as a result of mutations in the ribosome, a ubiquitous cellular machine, has remained a mystery. Combining mouse genetics and in vivo ribosome profiling
Melisa Herrero et al.
Cells, 10(8) (2021-08-28)
Translation initiation factor 2B (eIF2B) is a master regulator of global protein synthesis in all cell types. The mild genetic Eif2b5(R132H) mutation causes a slight reduction in eIF2B enzymatic activity which leads to abnormal composition of mitochondrial electron transfer chain
Kathrin Leppek et al.
Molecular cell, 80(6), 980-995 (2020-11-18)
Ribosomes have been suggested to directly control gene regulation, but regulatory roles for ribosomal RNA (rRNA) remain largely unexplored. Expansion segments (ESs) consist of multitudes of tentacle-like rRNA structures extending from the core ribosome in eukaryotes. ESs are remarkably variable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service